Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-9 Antibodies

          Invitrogen

          IL-9 Monoclonal Antibody (RM9A4), eFluor™ 660, eBioscience™

          9 Published Figures
          2 References
          View all (45) IL-9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-9 Monoclonal Antibody (RM9A4), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (9)
          IL-9 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-9 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-9 Antibody (50-8091-80) in Flow

          BALB/c splenocytes were cultured with plate-bound Anti-Mouse CD3e Functional Grade Purified (Product # 16-0031-82), Anti-Mouse CD28 Functional Grade Purified (Product # 16-0281-82), Mouse IL-4 Recombinant Protein (Product # 14-8041-80), and Human TGF beta 1 Recombinant Protein (Product # 14-8348-62) for 3 days, then cultured with Cell Stimulation Cocktail (plus protein transport inhibitors) (Product # 00-4975-03) for an additional 5 hours. Cells were stained intracellularly with Anti-Mouse CD4 FITC (Product # 11-0042-82) and 0.03 µg Anti-M... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)
          IL-9 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-8091-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.06 µg/test
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RM9A4

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11220480

          Product Specific Information

          Description: This monoclonal RM9A4 antibody reacts with mouse Interleukin-9 (IL-9). IL-9 is a 14 kDa cytokine originally named P40 and identified by its proliferative effects on T cell populations. The receptor, which is a heterodimer of the gamma chain portion of the IL-2 receptor and the IL-9R chain, activates Jak/STAT signaling pathways upon binding its ligand. Since the discovery of IL-9, numerous other functions have been observed. It induces Th17 and Treg differentiation in CD4+ T cells, IgE production in B cells, and the differentiation and proliferation of mast cells. IL-9 expression was initially observed in Th2 cells, but has since been found in Th17, eosinophil, and mast cells. Th9 cells, a newly discovered subset of CD4+ T cells, are characterized by the secretion of large amounts of IL-9 and IL-10. Th9 development is induced by stimulation of undifferentiated CD4+ with IL-4 and TGF beta. Th2 cells can also be driven towards a Th9 phenotype in the presence of TGF beta.

          Applications Reported: This RM9A4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This RM9A4 antibody has been tested by intracellular staining followed by flow cytometric analysis of cultured mouse splenocytes. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cytokine P40; ILN; Interleukin; Interleukin9; T-cell growth factor P40; unnamed protein product

          View more View less

          Gene Aliases: Il-9; P40

          View more View less

          UniProt ID: (Mouse) P15247

          View more View less

          Entrez Gene ID: (Mouse) 16198

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding interleukin-9 receptor binding growth factor activity
          Process(es)
          immune response immunoglobulin mediated immune response B cell differentiation positive regulation of cell growth positive regulation of interleukin-5 production interleukin-9-mediated signaling pathway B cell proliferation regulation of JAK-STAT cascade regulation of peptidyl-tyrosine phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch