Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Granzyme B Antibodies

          Invitrogen

          Granzyme B Monoclonal Antibody (NGZB), eFluor™ 660, eBioscience™

          44 Published Figures
          50 References
          View all (103) Granzyme B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Granzyme B Monoclonal Antibody (NGZB), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (44)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Granzyme B Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 45

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Granzyme B Antibody (50-8898-82) in Flow

          C57Bl/6 splenocytes were stimulated for 4 days with Anti-Mouse CD3e and Anti-Mouse CD28 Functional Grade Purified (Product # 16-0031-82 & Product # 16-0281-82) and Brefeldin A was added for the last 5 hours of culture. Cells were surface stained with Anti-Mouse CD8a FITC (Product # 11-0081-82) followed by intracellular staining with 0.06 µg of Rat IgG2a K Isotype Control eFluor® 660 (Product # 50-4321-82) (left) or 0.06 µg of Anti-Mouse Granzyme B eFluor® 660 (right) using the Intracellular Fixation & Permeabilization Buffer Set (plus Bref... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Immunohistochemistry (IHC)
          Granzyme B Antibody in Immunohistochemistry (IHC)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-8898-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 47 publications 47 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          NGZB

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • APC 
          • APC-eFluor 780
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11219679

          Product Specific Information

          Description: This NGZB monoclonal antibody reacts with mouse Granzyme B, which is a member of the granzyme serine protease family. Granzyme B is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. This NGZB monoclonal antibody does not cross-react to human Granzyme B nor is staining blocked with GB11, suggesting it recognizes a different epitope.

          Applications Reported: This NGZB antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This NGZB antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols®: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cytotoxic cell protease 1; Cytotoxic T lymphocyte associated serine esterase 1; fragmentin-2; GranzymeB; Gzmb; Human lymphocyte protein (Hlp); OTTHUMP00000028189; T-cell specific protein CCPI precursor; unnamed protein product

          View more View less

          Gene Aliases: CCP-1/C11; CCP1; Ctla-1; Ctla1; GZB

          View more View less

          UniProt ID: (Mouse) P04187

          View more View less

          Entrez Gene ID: (Mouse) 14939

          View more View less

          Function(s)
          serine-type endopeptidase activity protein binding serine-type peptidase activity
          Process(es)
          plasma membrane repair T cell mediated cytotoxicity positive regulation of immune response to tumor cell proteolysis negative regulation of translation protein catabolic process natural killer cell mediated cytotoxicity defense response to bacterium ceramide biosynthetic process protein maturation pyroptosis granzyme-mediated programmed cell death signaling pathway pyroptotic cell death
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-lpswf:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0