Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin Pan Type I/II Antibodies

          Invitrogen

          Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 660, eBioscience™

          Advanced Verification
          13 Published Figures
          16 References
          View all (75) Cytokeratin Pan Type I/II antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (13)
          • Advanced Verification (1)
          Pan Cytokeratin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Pan Cytokeratin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan Cytokeratin Antibody (50-9003-82) in ICC/IF

          Immunocytochemistry of fixed and permeabilized MCF7 cells using 5 µg/mL of Mouse IgG1 K Isotype Control eFluor® 660 (left) or 5 µg/mL of Anti-Pan Cytokeratin (AE1/AE3) eFluor® 660 (right). Nuclei are stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan Cytokeratin Antibody in Immunocytochemistry (ICC/IF)
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Cytokeratin Antibody in Western Blot (WB)
          Pan Cytokeratin Antibody in Immunocytochemistry (ICC/IF)
          Pan Cytokeratin Antibody in Immunocytochemistry (ICC/IF)
          Pan Cytokeratin Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Immunohistochemistry (IHC)
          Pan Cytokeratin Antibody in Western Blot (WB)
          Pan Cytokeratin Antibody in Flow Cytometry (Flow)
          Pan Cytokeratin Antibody in Flow Cytometry (Flow)
          Pan Cytokeratin Antibody in Flow Cytometry (Flow)
          Pan Cytokeratin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-9003-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate, Rabbit, Rhesus monkey

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AE1/AE3

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Alexa Fluor 420 
          • Alexa Fluor 488 
          • Alexa Fluor 514 
          • Alexa Fluor 700 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 750
          • eFluor 570
          • eFluor 615
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574301

          Product Specific Information

          Description: The monoclonal antibodies AE1 and AE3 recognize many of the acidic and basic cytokeratin family members. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. There are two large families of cytokeratins, acidic and basic, but all contain the same basic domains (i.e. an alpha-helical core with an N- and C-terminal domain). The proteins are expressed in epithelial cells, but are developmentally regulated. Many tumors also express these proteins and their expression can help identify the origin of a neoplasm.

          The AE3 monoclonal antibody recognizes the 65 to 67 triplet, 64, 59, 58, 56, 54 and 52kD proteins also known as cytokeratin 1, 2, 3, 4, 5, 6, 7, and 8 while the AE1 antibody recognizes 56.5, 54', 50, 50', 48, and 40 kDa proteins (also known as CK10, 14, 15, 16 and 19). These antibodies can be used on a wide array of tissue samples from mouse, human, rat, primates (cynomolgus and rhesus), dog, cat, rabbit, and chicken.

          Applications Reported: This AE1/AE3 antibody has been reported for use in immunohistochemical staining, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.

          Applications Tested: This AE1/AE3 antibody has been tested by immunocytochemistry on methanol-fixed MCF7 cells and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; 70 kd type II; cell proliferation-inducing gene 46 protein; CK-1; CK-10; CK-13; CK-15; CK-17; CK-19; CK-1B; CK-2e; CK-2P; CK-3; CK-4; CK-5; CK-6C; CK-6E; CK-7; CK-8; cytokeratin 1; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 2; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; cytokeratin endo A; cytokeratin endo B; Cytokeratin-1; cytokeratin-10; cytokeratin-15; cytokeratin-16; cytokeratin-17; cytokeratin-18; cytokeratin-1B; Cytokeratin-2e; cytokeratin-2P; cytokeratin-3; cytokeratin-4; cytokeratin-5; cytokeratin-6A; cytokeratin-6C; cytokeratin-6E; cytokeratin-7; cytokeratin-8; cytoplasmic intermediate filament protein; cytoskeletal 57 kDa keratin; cytoskeletal protein; embryonic type II keratin 1; EndoA cytokeratin (5' end put.); putative; EndoA' cytokeratin (5' end put.); putative; epidermal keratin 10; epidermal keratin type I; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; epithelial keratin 1; epithelial keratin 10; epithelial keratin 2e; epithelial keratin-2e; focal non-epidermolytic palmoplantar keratoderma; hair alpha protein; HLA-DRB1-restricted epitope S1; HLA-DRB1-restricted epitope S2; HLA-DRB1-restricted epitope S3; HLA-DRB1-restricted epitope S4; HMWCK; intermediate filament protein; intermediate filament protein K16; intermediate filament protein; K7; K1; K1B; K2e; K2P; K3 keratin; K6-alpha; K6C; K76; K77; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13; keratin 13, type I; keratin 14, type I; keratin 15, type I; keratin 16 (focal non-epidermolytic palmoplantar keratoderma); keratin 16 homolog; keratin 16, type I; keratin 17, type I; keratin 18, type I; keratin 19, type I; Keratin 1B; keratin 2 epidermis; keratin 2, type II; keratin 2A (epidermal ichthyosis bullosa of Siemens); keratin 2p; keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6A, , type II; keratin 6A, type II; keratin 6C, type II; keratin 6E; keratin 7, type II; keratin 76, type II; keratin 77; keratin 77, type II; keratin 8, type II; keratin 86; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 17; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 39; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin K3; keratin K5; keratin K6h; keratin type II; Keratin type II cytoskeletal 5; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; keratin-10; keratin-15, basic; keratin-15, beta; keratin-2 epidermis; keratin-2e; keratin-6-alpha; KRT1B; KRT5; masticatory epithelia keratin 2p; mK6-alpha; ORF; Pan CK; pan Cytokeratin; pan keratin; pankeratin; PIG46; sarcolectin; suprabasal cytokeratin 10; tuftelin interacting protein 8; type I cytoskeletal 15; type II cytoskeletal 1b; type II keratin Kb39; type II keratin subunit protein; type II mesothelial keratin K7; type-II intermediate filament protein; type-II keratin Kb1; type-II keratin Kb12; type-II keratin Kb2; Type-II keratin Kb3; Type-II Keratin Kb39; type-II keratin Kb4; type-II keratin Kb5; type-II keratin Kb6; type-II keratin Kb7; type-II keratin Kb8; type-II keratin Kb9; unnamed protein product

          View more View less

          Gene Aliases: 2310001L23Rik; 2310016L08Rik; 3300001P10Rik; 39.1; 4732484G22Rik; AEI2; BCIE; BIE; CARD2; CK-10; CK-14; CK-16; CK-17; CK-18; CK-19; CK-1B; CK-2e; CK-3; CK-4; CK-6A; CK-6C; CK-6E; CK-8; CK1; CK10; CK13; CK14; CK15; CK16; CK18; CK19; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK18; CYK4; CYK8; D130054E02Rik; D15Wsu77e; DDD; DDD1; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS2; EBS2A; EBS2B; EBS2C; EBS2D; EBS2E; EBS2F; EBS3; EBS4; EGK_03684; EGK_03685; EHK; EHK1; EHK2; EHK2A; EHK2B; EndoA; EndoC; EPPK; FNEPPK; HUMCYT2A; IHL; K1; K10; K13; K14; K15; K16; K17; K18; K19; K1B; K1C1; K1CO; K1CP; K1CS; K1emb; K2C7; K2C8; K2E; K3; K4; K5; K6[a]; K6A; K6C; K6D; K6E; K7; K76; K77; K8; KER1; Ker10; KO; KPP; Krt-1.10; Krt-1.13; Krt-1.14; Krt-1.19; Krt-2.1; Krt-2.4; Krt-2.8; Krt1-10; Krt1-13; Krt1-14; Krt1-15; Krt1-16; Krt1-17; Krt1-18; Krt1-19; KRT13; KRT16A; KRT1A; KRT1B; Krt2-1; Krt2-17; Krt2-2; Krt2-4; Krt2-5; Krt2-6a; Krt2-6c; Krt2-7; Krt2-8; Krt239; KRT2A; KRT2B; KRT2E; KRT2P; KRT5A; KRT6C; KRT6D; KRT6E; KRT77; Krt86; KRTE; MECD2; mK1b; mK6[a]; MK6a; NEPPK; NFJ; PC1; PC2; PC3; PCHC1; PPKNEFD; SCL; Tfip8; TROMA-1; WSN1; WSN2

          View more View less

          UniProt ID: (Dog) Q6EIY9, (Human) P04264, (Dog) Q6EIZ0, (Human) P13645, (Human) P13646, (Human) P02533, (Human) P19012, (Human) P08779, (Human) Q04695, (Dog) A0A8I3PJ84, (Human) P05783, (Human) P08727, (Human) P35908, (Rabbit) Q29426, (Human) P12035, (Human) P19013, (Human) P13647, (Human) P02538, (Human) P48668, (Human) P08729, (Human) Q01546, (Human) Q7Z794, (Human) P05787, (Mouse) P04104, (Mouse) P02535, (Mouse) P08730, (Mouse) Q61781, (Mouse) Q61414, (Mouse) Q9Z2K1, (Mouse) Q9QWL7, (Mouse) P05784, (Mouse) P19001, (Mouse) Q3TTY5, (Mouse) P07744, (Mouse) Q922U2, (Mouse) P50446, (Mouse) Q9DCV7, (Mouse) Q3UV17, (Mouse) Q6IFZ6, (Mouse) P11679

          View more View less

          Entrez Gene ID: (Dog) 444857, (Rhesus monkey) 699320, (Rabbit) 100353415, (Human) 3848, (Rabbit) 100009029, (Dog) 491006, (Human) 3858, (Rhesus monkey) 702153, (Human) 3860, (Rhesus monkey) 706830, (Dog) 490983, (Human) 3861, (Rhesus monkey) 100426180, (Dog) 100685323, (Rabbit) 100341120, (Rhesus monkey) 106994041, (Dog) 100685404, (Human) 3866, (Dog) 490978, (Rhesus monkey) 707236, (Human) 3868, (Rabbit) 100341376, (Rhesus monkey) 100426330, (Dog) 490977, (Human) 3872, (Rabbit) 100344942, (Dog) 477601, (Human) 3875, (Rabbit) 100354174, (Human) 3880, (Dog) 100685481, (Rabbit) 100353162, (Rhesus monkey) 699567, (Dog) 444850, (Human) 3849, (Dog) 100683401, (Rabbit) 100353668, (Rhesus monkey) 100425096, (Human) 3850, (Dog) 100682832, (Rhesus monkey) 700060, (Human) 3851, (Rabbit) 100348388, (Rhesus monkey) 100424400, (Rabbit) 100352658, (Dog) 100687760, (Human) 3852, (Dog) 486523, (Human) 3853, (Human) 286887, (Human) 3855, (Dog) 477602, (Rabbit) 100345579, (Rhesus monkey) 697425, (Rhesus monkey) 699194, (Dog) 486518, (Human) 51350, (Human) 374454, (Rabbit) 103348273, (Dog) 100683333, (Rabbit) 100348891, (Dog) 486513, (Rhesus monkey) 700449, (Human) 3856, (Mouse) 16678, (Mouse) 16661, (Mouse) 16663, (Mouse) 16664, (Mouse) 16665, (Mouse) 16666, (Mouse) 16667, (Mouse) 16668, (Mouse) 16669, (Mouse) 16681, (Mouse) 16682, (Mouse) 110308, (Mouse) 16687, (Mouse) 110310, (Mouse) 77055, (Mouse) 406220, (Mouse) 16691, (Rabbit) 100344183, (Rhesus monkey) 718983

          View more View less

          Function(s)
          protein heterodimerization activity protein binding carbohydrate binding structural constituent of epidermis signaling receptor activity structural molecule activity structural constituent of cytoskeleton keratin filament binding scaffold protein binding RNA binding structural constituent of muscle cadherin binding involved in cell-cell adhesion macromolecular complex binding cytoskeletal protein binding
          Process(es)
          keratinization intermediate filament cytoskeleton organization intermediate filament organization protein heterotetramerization morphogenesis of an epithelium cytoskeleton organization epithelial cell differentiation regulation of translation in response to stress wound healing negative regulation of keratinocyte differentiation keratinocyte development epidermis development keratinocyte differentiation positive regulation of epidermis development cornification cellular response to calcium ion tongue morphogenesis cellular response to retinoic acid Notch signaling pathway response to estrogen sarcomere organization cell differentiation involved in embryonic placenta development response to mechanical stimulus regulation of cell migration regulation of protein localization intermediate filament polymerization hair cycle intermediate filament bundle assembly positive regulation of keratinocyte differentiation positive regulation of cell growth hair follicle morphogenesis positive regulation of translation positive regulation of hair follicle development response to radiation positive regulation of cell proliferation cell differentiation negative regulation of cell migration killing of cells of other organism defense response to Gram-positive bacterium antimicrobial humoral immune response mediated by antimicrobial peptide negative regulation of entry of bacterium into host cell negative regulation of epithelial cell proliferation Wnt signaling pathway tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane protein transport negative regulation of apoptotic process extrinsic apoptotic signaling pathway hepatocyte apoptotic process complement activation, lectin pathway negative regulation of inflammatory response establishment of skin barrier response to hydrostatic pressure response to other organism epithelium development negative regulation of hepatocyte apoptotic process anatomical structure morphogenesis cell-cell adhesion pigmentation sebaceous gland development peptide cross-linking inflammatory response innate immune response keratinocyte migration keratinocyte activation keratinocyte proliferation negative regulation of epithelial cell differentiation response to oxidative stress fibrinolysis regulation of angiogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch