CD282 (TLR2) Monoclonal Antibody (6C2), eFluor™ 660, eBioscience™ - FAQs
CD282 (TLR2) Monoclonal Antibody (6C2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD282 (TLR2) Monoclonal Antibody (6C2), eFluor™ 660, eBioscience™ - FAQs (50-9021-80, 50-9021-82)