IL-17AF Monoclonal Antibody (B8KN8R), eFluor™ 660, eBioscience™ - FAQs
IL-17AF Monoclonal Antibody (B8KN8R), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17AF Monoclonal Antibody (B8KN8R), eFluor™ 660, eBioscience™ - FAQs (50-9171-80)