Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Leptin Antibodies

          Invitrogen

          Leptin Polyclonal Antibody, Biotin, PeproTech®

          View all (34) Leptin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Leptin Polyclonal Antibody, Biotin, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Leptin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Leptin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Leptin Antibody (500-P185GBT-25UG) in WB

          Western Blot: To detect Rat Leptin by Western Blot analysis Leptin Polyclonal Antibody, Biotin (Product # 500-P185GBT-500UG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat Leptin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in ELISA (ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Leptin Polyclonal Antibody, Biotin, PeproTech®

          Product Details

          500-P185GBT-25UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Rat

          Host/Isotype

          Goat

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Rat Leptin
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          25 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929712

          Product Specific Information

          AA Sequence of recombinant protein: MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC.

          Preparation: Produced from sera of goats immunized with highly pure Recombinant Rat Leptin. Anti-Rat Leptin-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect Rat Leptin by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat Leptin (500-P185G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat Leptin.

          Western Blot: To detect Rat Leptin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat Leptin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P185GBT-1MG will be provided as 2 x 500 µg

          Target Information

          Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD295 antigen; H-Leptin; HuB219; LEP-R; OB-R; obesity; obesity factor; obesity protein

          View more View less

          Gene Aliases: OB; obese

          View more View less

          UniProt ID: (Rat) P50596

          View more View less

          Entrez Gene ID: (Rat) 25608

          View more View less

          Function(s)
          DNA binding receptor binding cytokine activity hormone activity protein binding peptide hormone receptor binding leptin receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis ovulation from ovarian follicle response to hypoxia positive regulation of cytokine production placenta development regulation of endothelial cell proliferation response to dietary excess cardiac muscle hypertrophy glucose metabolic process regulation of gluconeogenesis energy reserve metabolic process glycerol biosynthetic process lipid metabolic process fatty acid beta-oxidation phagocytosis signal transduction JAK-STAT cascade female pregnancy response to nutrient circadian rhythm cholesterol metabolic process bile acid metabolic process regulation of blood pressure positive regulation of cell proliferation determination of adult lifespan adult feeding behavior fatty acid catabolic process negative regulation of autophagy negative regulation of lipid storage response to activity central nervous system neuron development insulin secretion T cell differentiation regulation of intestinal cholesterol absorption positive regulation of protein modification process response to nutrient levels positive regulation of TOR signaling negative regulation of appetite prostaglandin secretion response to estradiol positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production regulation of natural killer cell activation regulation of natural killer cell proliferation response to insulin cellular response to insulin stimulus response to vitamin E leptin-mediated signaling pathway positive regulation of luteinizing hormone secretion positive regulation of peroxisome proliferator activated receptor signaling pathway intracellular signal transduction bone mineralization involved in bone maturation aorta development negative regulation of appetite by leptin-mediated signaling pathway positive regulation of T cell proliferation regulation of natural killer cell mediated cytotoxicity positive regulation of protein import into nucleus hormone metabolic process glucose homeostasis eating behavior negative regulation of apoptotic process positive regulation of ion transport positive regulation of MAPK cascade cellular response to leptin stimulus response to ethanol regulation of fat cell differentiation regulation of angiogenesis negative regulation of vasoconstriction negative regulation of glucose import positive regulation of JAK-STAT cascade negative regulation of insulin receptor signaling pathway positive regulation of insulin receptor signaling pathway regulation of bone remodeling positive regulation of follicle-stimulating hormone secretion regulation of hormone secretion regulation of lipid biosynthetic process positive regulation of developmental growth regulation of catalytic activity regulation of insulin secretion regulation of steroid biosynthetic process intestinal absorption leukocyte tethering or rolling regulation of nitric-oxide synthase activity negative regulation of transport elastin metabolic process regulation of cell cycle positive regulation of protein kinase B signaling regulation of lipoprotein lipid oxidation adipose tissue development negative regulation of cartilage development negative regulation of glucagon secretion cellular response to L-ascorbic acid cellular response to retinoic acid regulation of brown fat cell differentiation energy homeostasis receptor signaling pathway via STAT bone growth positive regulation of cold-induced thermogenesis regulation of cytokine production involved in inflammatory response regulation of protein localization to nucleus positive regulation of p38MAPK cascade positive regulation of fat cell apoptotic process activation of protein kinase C activity positive regulation of reactive oxygen species metabolic process negative regulation of glutamine transport positive regulation of hepatic stellate cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0