Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-4 Antibodies

          Invitrogen

          IL-4 Polyclonal Antibody, PeproTech®

          2 Published Figures
          3 References
          View all (115) IL-4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-4 Polyclonal Antibody, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (2)
          IL-4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-4 Antibody (500-P94-100UG) in WB

          Western Blot: To detect Rat IL-4 by Western Blot analysis IL-4 Polyclonal Antibody (Product # 500-P94-1MG) can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat IL-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-4 Antibody in Western Blot (WB)
          IL-4 Antibody in Western Blot (WB)
          IL-4 Antibody in ELISA (ELISA)
          IL-4 Antibody in Neutralization (Neu)
          IL-4 Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          IL-4 Polyclonal Antibody, PeproTech®

          Product Details

          500-P94-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.5-2.0 µg/mL
          -

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Immunostaining (IS)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Rat

          Published species

          Rat

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Rat IL-4
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2930869

          Product Specific Information

          AA Sequence of recombinant protein: MHGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-4. Anti-Rat IL-4-specific antibody was purified by affinity chromatography employing an immobilized Rat IL-4 matrix.

          Sandwich ELISA: To detect Rat IL-4 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat IL-4 (500-P94Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-4.

          Western Blot: To detect Rat IL-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat IL-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B-cell growth factor 1; B-cell IGG differentiation factor; B-cell stimulatory factor 1; BSF-1; H-IL-4; IL-4; IL-4; cytokine; ILN; Interleukin; Interleukin4; lymphocyte stimulatory factor 1; M-IL-4; MGC79402

          View more View less

          Gene Aliases: Il4e12

          View more View less

          UniProt ID: (Rat) P20096

          View more View less

          Entrez Gene ID: (Rat) 287287

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding interleukin-4 receptor binding growth factor activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter microglial cell activation positive regulation of protein phosphorylation innate immune response in mucosa positive regulation of defense response to virus by host T-helper 1 cell lineage commitment positive regulation of immunoglobulin production negative regulation of acute inflammatory response negative regulation of chronic inflammatory response positive regulation of leukocyte chemotaxis negative regulation of leukocyte activation immune response JAK-STAT cascade female pregnancy response to nutrient cholesterol metabolic process positive regulation of cell proliferation response to xenobiotic stimulus response to nematode regulation of proton transport response to selenium ion positive regulation of gene expression negative regulation of epithelial cell migration positive regulation of macroautophagy positive regulation of cell migration positive regulation of B cell proliferation B cell costimulation negative regulation of tumor necrosis factor production positive regulation of chemokine production positive regulation of interleukin-10 production positive regulation of interleukin-13 production response to genistein response to cytokine interleukin-4-mediated signaling pathway positive regulation of T cell proliferation positive regulation of activated T cell proliferation T cell activation macrophage activation positive regulation of tyrosine phosphorylation of STAT protein defense response to protozoan myeloid dendritic cell differentiation negative regulation of macrophage activation positive regulation of mast cell degranulation negative regulation of nitric oxide biosynthetic process T-helper 2 cell differentiation leukocyte activation positive regulation of MHC class II biosynthetic process response to ethanol positive regulation of T cell differentiation negative regulation of osteoclast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade regulation of protein export from nucleus positive regulation of receptor-mediated endocytosis positive regulation of isotype switching to IgE isotypes positive regulation of isotype switching to IgG isotypes negative regulation of inflammatory response positive regulation of peptidyl-tyrosine phosphorylation regulation of immune response negative regulation of T cell activation positive regulation of B cell activation retina development in camera-type eye negative regulation of white fat cell proliferation cellular response to mercury ion positive regulation of mononuclear cell migration dendritic cell differentiation extrinsic apoptotic signaling pathway in absence of ligand positive regulation of cold-induced thermogenesis neuroinflammatory response positive regulation of beta-amyloid clearance positive regulation of myoblast fusion positive regulation of cellular respiration positive regulation of establishment of endothelial barrier positive regulation of reactive oxygen species biosynthetic process negative regulation of complement-dependent cytotoxicity negative regulation of cellular response to transforming growth factor beta stimulus negative regulation of T-helper 17 cell differentiation negative regulation of endothelial cell apoptotic process positive regulation of eosinophil chemotaxis positive regulation of B cell chemotaxis positive regulation of T-helper 2 cell cytokine production positive regulation of ATP biosynthetic process negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0