Search
View additional product information for Clear Aid - FAQs (5005)
1 product FAQs found
An alternative for PureCol, Type I Collagen Solution, 3 mg/ml (Bovine) (Cat. No. 5005) would be Collagen I, bovine (Cat. No. A1064401), which can be found here:
Collagen I, bovine (Cat. No. A1064401)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.