GPR109 Monoclonal Antibody (4NZBRGO), Alexa Fluor™ 647, eBioscience™ - FAQs
GPR109 Monoclonal Antibody (4NZBRGO), Alexa Fluor™ 647, eBioscience™ - FAQs
View additional product information for GPR109 Monoclonal Antibody (4NZBRGO), Alexa Fluor™ 647, eBioscience™ - FAQs (51-4779-42)