Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • 14-3-3 Pan Antibodies

          Invitrogen

          14-3-3 Pan Polyclonal Antibody

          Advanced Verification
          12 Published Figures
          14 References
          View all (7) 14-3-3 Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite 14-3-3 Pan Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (12)
          • Advanced Verification (1)
          14-3-3 Pan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          14-3-3 Pan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          14-3-3 Pan Antibody (51-0700) in ICC/IF

          Immunofluorescence analysis of 14-3-3 theta was performed using 70% confluent log phase MCF-7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with 14-3-3 theta Rabbit Polyclonal Antibody (Product # 51-0700) at 2 µg/mL in 0.1% BSA and incubated for 3 hours ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          14-3-3 Pan Antibody in Immunocytochemistry (ICC/IF)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Immunohistochemistry (IHC)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody in Western Blot (WB)
          14-3-3 Pan Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          14-3-3 Pan Polyclonal Antibody

          Product Details

          51-0700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 2 publications 2 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat, Xenopus

          Published species

          Bovine, Fruit fly, Human, Mouse, Plant

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A 20 amino acid synthetic peptide derived from the N-terminus of the human 14-3-3 beta/alpha protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          200 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533894

          Product Specific Information

          This antibody is broadly reactive with members of the 14-3-3 family of proteins (~29-32 kDa). With the exception of a single conserved amino acid change, this sequence is identical in the delta and zeta isoforms. Three and four conserved amino acid differences are found in the tau and gamma isoforms, respectively.

          Based on the sequence homology to the immunizing peptide, this the antibody could recognize the protein from C. elegans.

          Target Information

          At least seven isoforms comprise the highly conserved 14-3-3 family of homo- and heterodimeric proteins that are abundantly expressed in all eukaryotic cells. Although more than seven isoforms of 14-3-3 have been described, some redundancies have appeared upon sequencing. The 14-3-3s are thought to be key regulators of signal transduction events mediated through their binding to serine-phosphorylated proteins. By interacting with Cdc25C, 14-3-3 regulates entry into the cell cycle and, through interaction with Bad, prevents apoptosis. Other proteins that have been shown to bind to 14-3-3s include members of the protein kinase C family, Cbl, IRS-1, polyoma middle-T antigen, nitrate reductase, S-raf and the IGF-1 receptor. Detection of 14-3-3 proteins in cerebrospinal fluid has been shown to be quite useful in the differential diagnosis of Creutzfeldt-Jakob disease and other prion diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 14-3-3 alpha; 14-3-3 beta; 14-3-3 delta; 14-3-3 epsilon; 14-3-3 gamma; 14-3-3 protein beta; 14-3-3 protein beta-subtype; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha-A; 14-3-3 protein gamma subtype; 14-3-3 protein gamma-subtype; 14-3-3 protein T-cell; 14-3-3 protein tau; 14-3-3 protein zeta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 sigma protein; 14-3-3 tau; 14-3-3 theta; 14-3-3 zeta; 14-3-3d; 14-3-3E; 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; 3-monooxgenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; epididymis luminal protein 2; epididymis luminal protein 4; epididymis secretory protein Li 1; epididymis secretory protein Li 3; epididymis secretory protein Li 93; epithelial cell marker protein 1; factor activating exoenzyme S; FAS; HS1; KCIP-1; mitochondrial import stimulation factor (MSF) L subunit; mitochondrial import stimulation factor L subunit; mitochondrial import stimulation factor S1 subunit; MME1; MSF L; phospholipase A2; prepronerve growth factor homolog; prepronerve growth factor RNH-1; protein 1054; protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; protein phosphatase 1, regulatory subunit 170; protein tau; protein, theta; putative protein kinase C regulatory protein; SEZ-2; SFN; signal transduction phosphoprotein; tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma; tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alpha polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, delta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activatioprotein epsilon polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activatioprotein, epsilon polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, theta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; unnamed protein product; YWHAG

          View more View less

          Gene Aliases: 1110013I11Rik; 1300003C17Rik; 14-3-3; 14-3-3-zeta; 14-3-3b; 14-3-3beta; 14-3-3E; 14-3-3G; 14-3-3GAMMA; 14-3-3t; 14-3-3z; 14-3-3zeta; 1C5; 2700028P07Rik; D7Bwg1348e; DEE56; EIEE56; Er; GW128; HEL-S-1; HEL-S-3; HEL-S-93; HEL2; HEL4; HS1; KCIP-1; MDCR; MDS; Mme1; POPCHAS; PPP1R170; XELAEV_18012695mg; XELAEV_18015318mg; XELAEV_18043238mg; XELAEV_18045999mg; YWHAA; ywhab; ywhab-a; ywhab-b; YWHAD; ywhag; ywhag-a; ywhag-b; YWHAS

          View more View less

          UniProt ID: (Bovine) Q0VC36, (Human) P31947, (Mouse) O70456, (Bovine) P68250, (Human) P31946, (Bovine) P62261, (Human) P62258, (Human) P61981, (Bovine) Q3SZI4, (Human) P27348, (Human) P63104, (Bovine) P63103, (Rat) P35213, (Mouse) Q9CQV8, (Mouse) P62259, (Rat) P62260, (Rat) P61983, (Mouse) P61982, (Rat) P68255, (Mouse) P68254, (Rat) P63102, (Mouse) P63101, (Xenopus) Q5XHK2, (Xenopus) Q8AVQ3, (Xenopus) Q6PCG0, (Xenopus) Q6NRY9

          View more View less

          Entrez Gene ID: (Bovine) 528453, (Human) 2810, (Rat) 313017, (Mouse) 55948, (Bovine) 286863, (Human) 7529, (Bovine) 282125, (Human) 7531, (Human) 7532, (Bovine) 286862, (Bovine) 768311, (Human) 10971, (Human) 7534, (Bovine) 287022, (Rat) 56011, (Mouse) 54401, (Mouse) 22627, (Rat) 29753, (Rat) 56010, (Mouse) 22628, (Rat) 25577, (Mouse) 22630, (Rat) 25578, (Mouse) 22631, (Xenopus) 399276, (Xenopus) 380457, (Xenopus) 379724, (Xenopus) 432005, (Xenopus) 100505434

          View more View less

          Function(s)
          protein kinase binding identical protein binding phosphoprotein binding protein binding protein kinase C inhibitor activity cadherin binding phosphoserine binding protein sequestering activity protein domain specific binding protein kinase inhibitor activity protein phosphatase inhibitor activity enzyme binding histone deacetylase binding macromolecular complex binding RNA binding calcium channel regulator activity potassium channel regulator activity calcium channel inhibitor activity protein phosphatase binding MHC class II protein complex binding ubiquitin protein ligase binding signaling adaptor activity ion channel binding protein heterodimerization activity scaffold protein binding molecular function inhibitor activity protein kinase C binding insulin-like growth factor receptor binding receptor tyrosine kinase binding phosphorylation-dependent protein binding 14-3-3 protein binding DNA-binding transcription factor binding enzyme inhibitor activity actin binding enzyme regulator activity scaffold/adaptor protein
          Process(es)
          regulation of cyclin-dependent protein serine/threonine kinase activity release of cytochrome c from mitochondria keratinocyte development intrinsic apoptotic signaling pathway in response to DNA damage regulation of epidermal cell division negative regulation of keratinocyte proliferation positive regulation of cell growth keratinization negative regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of epidermal cell differentiation positive regulation of protein export from nucleus establishment of skin barrier negative regulation of protein kinase activity protein export from nucleus signal transduction protein localization regulation of cell-cell adhesion keratinocyte differentiation regulation of protein localization positive regulation of hippo signaling positive regulation of keratinocyte differentiation positive regulation of cell adhesion negative regulation of innate immune response cAMP/PKA signal transduction negative regulation of protein localization to plasma membrane positive regulation of cellular protein localization biological_process skin development regulation of cell cycle defense response to bacterium establishment of protein localization cytolysis in other organism non-canonical inflammasome complex assembly protein targeting negative regulation of G-protein coupled receptor protein signaling pathway negative regulation of protein import into nucleus negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter MAPK cascade protein localization to nucleus cellular response to heat cytoplasmic pattern recognition receptor signaling pathway regulation of heart rate by hormone regulation of mitotic cell cycle substantia nigra development cellular potassium ion homeostasis negative regulation of toll-like receptor signaling pathway intracellular signal transduction regulation of cytosolic calcium ion concentration regulation of membrane repolarization protein localization to endoplasmic reticulum membrane repolarization during cardiac muscle cell action potential regulation of heart rate by cardiac conduction regulation of potassium ion transmembrane transport negative regulation of calcium ion export across plasma membrane positive regulation of T cell mediated immune response to tumor cell regulation of signal transduction positive regulation of cell-cell adhesion cellular response to insulin stimulus cellular response to glucose starvation regulation of neuron differentiation regulation of synaptic plasticity positive regulation of T cell activation negative regulation of TORC1 signaling negative regulation of ion transmembrane transport negative regulation of transcription from RNA polymerase II promoter angiogenesis respiratory system process protein phosphorylation synaptic target recognition lung development regulation of protein stability tube formation negative regulation of apoptotic process establishment of Golgi localization regulation of ERK1 and ERK2 cascade regulation of synapse maturation Golgi reassembly negative regulation of protein localization to nucleus negative regulation of cell growth involved in contact inhibition neuron migration ER to Golgi vesicle-mediated transport Golgi to plasma membrane transport hippocampus development cerebral cortex development positive regulation of toll-like receptor signaling pathway toll-like receptor 4 signaling pathway hippo signaling protein K11-linked ubiquitination regulation of postsynaptic membrane neurotransmitter receptor levels cytoplasmic translation negative regulation of T cell mediated immune response to tumor cell SCF-dependent proteasomal ubiquitin-dependent protein catabolic process cellular response to nutrient levels cellular response to amino acid starvation TORC1 signaling proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of translational initiation positive regulation of translational initiation protein stabilization negative regulation of T cell activation protein localization to lysosome protein K6-linked ubiquitination positive regulation of TORC1 signaling small GTPase mediated signal transduction histamine secretion by mast cell response to xenobiotic stimulus regulation of programmed cell death ERK1 and ERK2 cascade protein localization to mitochondrion lysosome organization antibacterial innate immune response regulation of lysosome organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch