Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Connexin 26 Antibodies

          Invitrogen

          Connexin 26 Polyclonal Antibody

          73 Published Figures
          93 References
          View all (8) Connexin 26 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Connexin 26 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (73)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 77

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 26 Antibody (51-2800) in ICC/IF

          Immunofluorescence analysis of Connexin 26/GJB2 was done on 70% confluent log phase Caco-2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with Connexin 26/GJB2 Rabbit polyclonal Antibody (Product # 51-2800) at 2 µg/mL in 1% BSA and incubated for 3 hours ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 26 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Connexin 26 Polyclonal Antibody

          Product Details

          51-2800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 27 publications 27 publications

          Immunohistochemistry (IHC)

          -
          View 34 publications 34 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          10-20 µg/mL
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          View 20 publications 20 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A 13 amino acid synthetic peptide derived from the C-terminus of the mouse Connexin 26 protein. This mouse sequence differs from the rat sequence by a single amino acid and from the human sequence by two (non-consecutive) amino acids.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533903

          Target Information

          Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: connexin 26; connexin e; connexin-26; connexin26; transmembrane protein; mutant CX26 protein; gap junction beta 2 proteinc; gap junction channel protein connexin 26; gap junction membrane channel protein beta 2; gap junction protein; gap junction protein, beta 2, 26kDa; mutant connexin 26; transmembrane protein; mutant gap junction beta 2 protein; mutant gap junction protein beta 2; unnamed protein product

          View more View less

          Gene Aliases: BAPS; Cnx26; CX26; CXN-26; Cxne; DFNA3; DFNA3A; DFNB1; DFNB1A; Gjb-2; HID; KID; NSRD1; PPK

          View more View less

          UniProt ID: (Human) P29033, (Rat) P21994, (Mouse) Q00977

          View more View less

          Entrez Gene ID: (Human) 2706, (Rat) 394266, (Mouse) 14619

          View more View less

          Function(s)
          gap junction channel activity calcium ion binding protein binding identical protein binding gap junction channel activity involved in cell communication by electrical coupling
          Process(es)
          response to ischemia cell communication cell-cell signaling female pregnancy cell communication by electrical coupling gap junction assembly response to estradiol response to lipopolysaccharide response to retinoic acid response to progesterone cellular response to oxidative stress response to human chorionic gonadotropin decidualization inner ear development transmembrane transport cellular response to glucagon stimulus cellular response to dexamethasone stimulus epididymis development gap junction-mediated intercellular transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-qh5rm:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0