Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 2 Antibodies

          Invitrogen

          Claudin 2 Polyclonal Antibody (MH44)

          Advanced Verification
          72 Published Figures
          158 References
          View all (12) Claudin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 2 Polyclonal Antibody (MH44)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (72)
          • Advanced Verification (3)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 78

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 2 Antibody (51-6100) in ICC/IF

          Immunofluorescence analysis of Claudin-2 was performed using 70% confluent log phase 769-P cells. The cells were fixed with 4% paraformaldehyde for 5 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Claudin 2 Polyclonal Antibody (MH44) (Product # 51-6100) at 1:100 dilution in 0.1% BSA, incubated at 4 degree... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 2 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 2 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          Claudin 2 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
          Claudin 2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 2 Antibody in Immunohistochemistry (IHC)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody in Western Blot (WB)
          Claudin 2 Antibody
          Claudin 2 Antibody
          Claudin 2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Claudin 2 Polyclonal Antibody (MH44)

          Product Details

          51-6100

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.25 - 2 µg/mL
          View 72 publications 72 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 25 publications 25 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:100
          View 45 publications 45 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.1-1.0 µg/mL
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Rat

          Published species

          Dog, Fish, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Rodent, Sheep

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Clone

          MH44

          Immunogen

          Synthetic peptide derived from the C-terminus of the human Claudin-2 protein
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533911

          Product Specific Information

          This antibody is specific for the ~ 22 kDa Claudin-2 protein and does not appear to cross-react with related endogenous proteins. Reactivity was confirmed by western blotting with rat kidney and by immunostaining of rat small intestine. This antibody is confirmed reactive with human, rat, and dog Claudin-2. The reactivity of this antibody with other species has not been determined.

          Target Information

          Tight junctions are specialized regions of cell-cell contact that are particularly abundant in luminal epithelial cell sheets. In freeze-fracture electron micrographs, tight junctions are visualized as belt-like bands of anastomosing sealing strands (TJ strands) that completely encircle the lateral surfaces of each cell. TJ strands on adjacent cells are presumed to interact with each other to form a sort of "molecular gasket" that prevents ions, water and other molecules from leaking between cells and thus, from one side of the sheet to the other. In addition to this so-called "barrier" function, the "fence" function of tight junctions plays an important role in maintaining epithelial cell-polarity by blocking the diffusion of membrane proteins between apical (luminal) and basolateral cell surfaces. Confinement of, for example, the glucose symport to apical surfaces allows glucose to be transported vectorially from the lumen, through the cell, and into the bloodstream. Several peripheral membrane proteins are associated with tight junctions including ZO-1, ZO-2, ZO-3 (members of membrane-associated guanylate-kinase family), cingulin, the 7H6 antigen, Rab-3b, symplekin. While their precise functions are not known, roles for these proteins have been suggested in tight junction assembly and maintenance; signal transduction; and the regulation of tight junction permeability. Furthermore, a growing body of evidence suggests that actin filaments play a major role in regulating tight junction permeability. Until recently, the only transmembrane protein known to be associated with tight junctions was occludin, an ~65 kDa protein with four transmembrane domains. Despite widespread expectation, a critical structural role for occludin in TJ strands was ruled out by the observation of apparently normal tight junctions formed between cells disrupted at both occludin alleles. Fortunately, a closer examination of isolated tight junctions uncovered two related ~22 kDa, four-transmembrane domain proteins, claudin-1 and claudin-2, with no similarity to occludin. In contrast to occludin, which induces only a small number of short strands at cell-cell contact sites when introduced into fibroblasts lacking tight junctions, claudin-1 and -2 induce networks of strands characteristic of true tight junctions. Though inconclusive, these findings suggest that claudin-1 and -2 are major structural components of TJ strands and that occludin plays some other accessory role. Excitement in the tight junction field continues to rise following the recent discovery of claudins -3, -4, -5, -6, -7, and -8 and experiments suggesting that tight junctions in different tissues are comprised of different sets of claudin family proteins. Claudin-1 and claudin-2 connect to the actin cytoskeleton through ZO-1; Claudin-2 functions as a paracellular channel with cation (Na+) selectivity at tight junctions. The expression of claudin-2 is restricted to the liver and kidney, with small amounts also found in the brain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Claudin2; CLD2; integral membrane protein claudin-2; SP82; tight junction-associated protein; unnamed protein product

          View more View less

          Gene Aliases: claudin-2; OAZON; RGD1560247

          View more View less

          UniProt ID: (Human) P57739, (Dog) Q95KM6

          View more View less

          Entrez Gene ID: (Human) 9075, (Dog) 403649, (Rat) 300920

          View more View less

          Function(s)
          structural molecule activity protein binding identical protein binding paracellular tight junction channel activity tight junction
          Process(es)
          innate immune response in mucosa calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules cell-cell adhesion regulation of bile acid secretion paracellular transport regulation of intestinal D-glucose absorption regulation of intestinal lipid absorption
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch