Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Connexin 36 Antibodies

          Invitrogen

          Connexin 36 Polyclonal Antibody

          84 Published Figures
          55 References
          View all (13) Connexin 36 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Connexin 36 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (84)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 84

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 36 Antibody (51-6300) in ICC/IF

          PloS one 2019 - Fig 1 Immunofluorescence staining of MAP2 in PC12 cells and colocalization of Cx36 and CaMKII in PC12 cells. (A) PC12 cells exhibited neuron-like characteristics. MAP2-positive staining was in red, and the nuclei were stained with DAPI in blue. (B) Colocalization of Cx36 and CaMKII in PC12 cells. The positive Cx36 staining was in green and CaMKII was in red. The nuclei were in blue. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Immunoprecipitation (IP)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Connexin 36 Polyclonal Antibody

          Product Details

          51-6300

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 22 publications 22 publications

          Immunohistochemistry (IHC)

          -
          View 15 publications 15 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 9 publications 9 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -

          Immunoprecipitation (IP)

          2-3 µg
          View 3 publications 3 publications

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Fish, Human, Mouse, Rat, Rodent

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide corresponding to a sequence from the C-terminus of rat and mouse Connexin 36 (Cx36). This sequence differs from the human Cx36 protein by a single amino acid
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533913

          Target Information

          Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: connexin s; connexin-36; gap junction alpha-9 protein; gap junction membrane channel protein alpha 9; gap junction protein, alpha 9; gap junction protein, delta 2, 36kDa; GJA10

          View more View less

          Gene Aliases: connexin36; CX36; Cxns; GJA9

          View more View less

          UniProt ID: (Human) Q9UKL4, (Rat) O70610, (Mouse) O54851

          View more View less

          Entrez Gene ID: (Human) 57369, (Rat) 50564, (Mouse) 14617

          View more View less

          Function(s)
          gap junction channel activity protein binding
          Process(es)
          cell communication cell-cell signaling transmembrane transport action potential chemical synaptic transmission visual perception
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-qh5rm:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0