Pierce™ Protein A/G UltraLink™ Resin
Thermo Scientific™

Pierce™ Protein A/G UltraLink™ Resin

Thermo Scientific Pierce Protein A/G UltraLink Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein A/G hasRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
531322 mL
5313310 mL
Catalog number 53132
Price (USD)
469.65
Online exclusive
528.00
Save 58.35
Each
Quantity:
2 mL
Request bulk or custom format
Price (USD)
469.65
Online exclusive
528.00
Save 58.35
Each
Thermo Scientific Pierce Protein A/G UltraLink Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein A/G has been immobilized for use in IgG purification methods.

Features of Protein A/G UltraLink Resin:

• Immobilized Protein A/G – specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
• Standard capacity – loading of immobilized Protein A/G provides a binding capacity of approx. 20 mg human IgG per milliliter of resin
• UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Protein A/G UltraLink Resin is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein A/G is a recombinant fusion protein consisting of the antibody-binding domains of Protein A and Protein G, enabling antibody purification from serum samples of a variety of species.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Properties of Thermo Scientific UltraLink Resin:
• Support pH Stability: 1 to 13
• Average Particle Size: 50 to 80 microns
• Exclusion Limit: greater than 2,000,000 daltons
• Average Surface Area: greater than 250m2 per gram of beads
• Average Pore Volume: greater than 1.2 mL per gram of beads (> 60% of bead volume)
• Pore Size: 500 to 1000 angstroms (50 to 100nm)
• Maximum Linear Velocity: 3000 cm per hour
• Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Related Products
Pierce™ Protein A/G Plus UltraLink™ Resin
Pierce™ Protein A/G Agarose
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Purification TargetIgG Antibodies
Quantity2 mL
Column TypeUltraLink Affinity Resin
FormatBottle
Product LinePierce, UltraLink
Product TypeResin
Stationary PhaseProtein A/G
Unit SizeEach
Contents & Storage
Upon receipt store resin at 4°C. Product is shipped at ambient temperature.