UltraLink™ Iodoacetyl Resin
Thermo Scientific™

UltraLink™ Iodoacetyl Resin

Thermo Scientific UltraLink Iodoacetyl Resin is a durable, porous resin that is activated to enable efficient and direct covalent immoblizationRead more
Have Questions?
Catalog NumberQuantity
5315510 mL
Catalog number 53155
Price (USD)
365.00
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
365.00
Each
Thermo Scientific UltraLink Iodoacetyl Resin is a durable, porous resin that is activated to enable efficient and direct covalent immoblization of peptides and other ligands through their sulfhydryl groups (-SH) for use in affinity purification procedures.

Features of UltraLink Iodoacetyl Resin:

• High coupling efficiency and capacity—immobilizes sulfhydryl-containing proteins or other ligands with high efficiency and coupling capacity in 1 hour
• Specific and leak-proof coupling chemistry—reacts specifically with sulfhydryl groups (reduced thiols), resulting in thioether bonds that are stable for use in many affinity purification procedures
• Simple coupling conditions—perform immobilization reaction in any of a variety of buffers; coupling is most efficient and specific at pH 8.0-8.5; coupling compatible with or without organic solvent
• Excellent reusability—prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification without significant loss of binding capacity
• Durable, high-performance resin—porous beads have a 60 μM diameter, can withstand 100 psi (6.9 bar) and allow for linear flow-through rates of 3,000 cm/hour

The beaded resin is a hydrophilic copolymer of polyacrylamide and azlactone having a rigid polymeric structure with high surface area and pore volume. Each azlactone group has been modified to form a 15-atom spacer arm that terminates in an iodoacetyl group, which is capable of reacting with sulfhydryl groups (e.g., side chain of reduced cysteine residues) to covalently immobilize peptide or other sulfhydryl-containing ligands. The bead structure and efficiently coupling chemistry of Iodoacetyl-Activated UltraLink Support results in high protein binding capacity, high linear flow rates, low nonspecific binding and overall superior performance in affinity chromatography. UltraLink Resins are ideal for medium pressure applications such as FPLC.

Properties of UltraLink Iodoacetyl Resin:
• Alternate names: 3M Emphaze Biosupport Medium AB1
• Matrix: Polyacrylamide/azlactone copolymer derivatized with iodoacetyl groups
• Particle size: 50 to 80 μM diameter
• Maximum pressure: 100 psi (6.9 bar)
• Maximum linear flow rate: 3000 cm/hour
• Coupling capacity: > 6 mg of reduced human IgG per ml of settled resin
• Leaving group in coupling reaction: Hydrogen iodide
• Bond formed: Thioether bond
• Spacer arm: 15 atoms

Related Products
UltraLink™ Iodoacetyl Micro Peptide Coupling Kit, 0.1 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, UltraLink™ Resin
DescriptionUltraLink Iodoacetyl Resin
FormatBottle
Quantity10 mL
Stationary PhaseHaloacetyl
Binding Capacity≥6 mg Human IgG Coupling/mL Resin
Particle Size50 to 80 μm
Product LineUltraLink
TypeResin
Unit SizeEach
Contents & Storage
Storage: Upon receipt store product at 4°C, protected from light. Product is shipped at ambient temperature