Axiom™ Maize6H Genotyping Array (60K), 384HT format
Applied Biosystems™

Axiom™ Maize6H Genotyping Array (60K), 384HT format

The Axiom Maize6H Genotyping Array (60K) is a component of our Axiom Genotyping Solution in which target genomic DNA isRead more
Have Questions?
Catalog number 551098
Price (USD)
-
The Axiom Maize6H Genotyping Array (60K) is a component of our Axiom Genotyping Solution in which target genomic DNA is prepared and processed on the GeneTitan Multi-Channel Instrument. The automated process includes hybridization of the gDNA to this high-throughput microarray, followed by staining, washing, imaging, and output of SNP genotyping data for further analysis. The Axiom Maize6H Genotyping Array (60K) was developed and designed in collaboration with the Maize Research Center, Beijing Academy of Agriculture and Forestry Sciences, China. This array is also available in mini-96 format (Cat. No. 551099).

Axiom Maize6H Genotyping Array (60K) content
• Contains 61,282 markers that are evenly distributed across the entire genome
• 90% of the content is classified as high-performance markers with MAF and PIC >0.2
• Based on the whole genome sequencing data of 388 representative maize inbred lines that were filtered for high SNP quality scores
• 100 chloroplast loci present

Applications
• Construction of DNA fingerprints of maize varieties and identification of similar varieties
• Gene mapping and genome-wide selection
• Molecular marker-assisted selection breeding
• Population genetics and evolution
• Evaluation of germplasm resources at a genome level

Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software
Axiom Long Format Export Tool (AxLE)

Reference

1. Tian, H.L., Yang, Y., Yi, H.M., Xu, L.W., He, H., Fan, Y.M., Wang, L., Ge, G.R., Liu, Y.W., Wang, F.G. and Zhao, J.R. (2021) New resources for genetic studies in maize (Zea mays L.): a genome-wide Maize6H-60K SNP array and its application. The Plant Journal 105, 1113-1122.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 array
Shipping ConditionRoom temperature
ArrayGenotyping
Format384 HT-array Plate
Number of Arrays384 arrays
SpeciesMaize
Unit Size1 array