HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Thermo Scientific™
HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
60104-23448 Position Extraction Plate Plugs
60104-23524 Position Extraction Plate Plugs
Catalog number 60104-234
Price (USD)
44.56
Each
Type:
48 Position Extraction Plate Plugs
Price (USD)
44.56
Each
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
Specifications
Description48 Position Extraction Plate Plugs
For Use With (Application)SPE Sample Processing
For Use With (Equipment)48 Position Well Extraction Plates, Vacuum Manifolds
Product LineHyperSep
Type48 Position Extraction Plate Plugs
Unit SizeEach