CD115 (c-fms) Monoclonal Antibody (AFS98), PE-eFluor™ 610, eBioscience™ - FAQs
CD115 (c-fms) Monoclonal Antibody (AFS98), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD115 (c-fms) Monoclonal Antibody (AFS98), PE-eFluor™ 610, eBioscience™ - FAQs (61-1152-82)