ROR gamma (t) Monoclonal Antibody (B2D), PE-eFluor™ 610, eBioscience™ - FAQs
ROR gamma (t) Monoclonal Antibody (B2D), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for ROR gamma (t) Monoclonal Antibody (B2D), PE-eFluor™ 610, eBioscience™ - FAQs (61-6981-82, 61-6981-80)