TNF alpha Monoclonal Antibody (MAb11), PE-eFluor™ 610, eBioscience™ - FAQs
TNF alpha Monoclonal Antibody (MAb11), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for TNF alpha Monoclonal Antibody (MAb11), PE-eFluor™ 610, eBioscience™ - FAQs (61-7349-41, 61-7349-42)