RPMI 1640 Medium, GlutaMAX™ Supplement, 10 x 500 mL - FAQs
RPMI 1640 Medium, GlutaMAX™ Supplement, 10 x 500 mL - FAQs
View additional product information for RPMI 1640 Medium, GlutaMAX™ Supplement - FAQs (61870044)
4 product FAQs found
How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?
While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the density (g/L) for RPMI 1640 Medium?
We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?
In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.