Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD11c Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          CD11c Monoclonal Antibody (N418), Super Bright™ 436, eBioscience™

          54 Published Figures
          59 References
          View all (196) CD11c antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols

          Cite CD11c Monoclonal Antibody (N418), Super Bright™ 436, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (54)
          CD11c Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD11c Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 55

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD11c Antibody (62-0114-80) in Flow

          C57BL/6 mouse splenocytes were stained with CD45R (B220) Monoclonal Antibody, FITC (Product # 11-0452-82) and 0.25 µg of Armenian Hamster IgG Isotype Control, Super Bright 436 (Product # 62-4888-82) (left) or 0.25 µg of CD11c Monoclonal Antibody, Super Bright 436 (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Immunohistochemistry (IHC)
          CD11c Antibody in Immunocytochemistry (ICC/IF)
          CD11c Antibody in Immunocytochemistry (ICC/IF)
          CD11c Antibody in Immunohistochemistry (IHC)
          CD11c Antibody in Immunohistochemistry (IHC)
          CD11c Antibody in Immunohistochemistry (IHC)
          CD11c Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD11c Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD11c Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD11c Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD11c Antibody in Immunohistochemistry (IHC)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)
          CD11c Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          62-0114-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 49 publications 49 publications
          Product Specifications

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), Super Bright™ 436, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N418

          Conjugate

          Super Bright™ 436 Super Bright™ 436 Super Bright™ 436
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • eFluor 450
          • eFluor 506
          • eFluor 615
          • eFluor 660
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 610-30S
          • NovaFluor Blue 660-40S
          • NovaFluor Blue 690
          • NovaFluor Blue 725
          • NovaFluor Blue 800
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 800
          • NovaFluor Yellow 570
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 600
          • Super Bright 645
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          413/431 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_2722927

          Product Specific Information

          Description: The N418 monoclonal antibody reacts with mouse CD11c, the integrin alpha X. CD11c non-covalently associates with beta 2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90 kDa heterodimer.

          Applications Reported: This N418 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This N418 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch