In-Cell ELISA Kit, Colorimetric - FAQs
In-Cell ELISA Kit, Colorimetric - FAQs
View additional product information for In-Cell ELISA Kit, Colorimetric - FAQs (62200)
1 product FAQs found
Do you offer alternatives for discontinued ELISPOT kits?
If you are unable to find the ELISPOT kit for your target protein, please consider Pierce Colorimetric In-Cell ELISA Kit (Cat. No. 62200). Please note that this format does not include primary antibodies and you will need to choose the antibody pair for the targets you wish to detect. However, The HRP Conjugate included in each kit will recognize and bind primary antibodies (IgG) from several species, including mouse and rabbit IgG and is therefore quite flexible.