UV Transilluminator Bulb, 306 nm, for myECL™ Imager - FAQs
UV Transilluminator Bulb, 306 nm, for myECL™ Imager - FAQs
View additional product information for UV Transilluminator Bulb, 306 nm, for myECL™ Imager - FAQs (62258)
2 product FAQs found
Do you offer any alternatives for the discontinued UV Transilluminator Bulb, 365 nm, for myECL Imager (Cat. No. 62255)?
No, we do not offer any alternatives for UV Transilluminator Bulb, 365 nm, for myECL Imager.
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .
Do you offer any alternatives for the discontinued UV Transilluminator Bulb, 306 nm, for myECL Imager (Cat. No. 62258)?
No, we do not offer any alternatives for UV Transilluminator Bulb, 306 nm, for myECL Imager.
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .