I have the Thermo Scientific G2 Fast Blotter (Cat. No. 62288). Has this device been discontinued? Can I get the 1-Step Transfer Buffer recommended for fast transfer with the Thermo Scientific G2 Fast Blotter?
Yes. The Thermo Scientific G2 Fast Blotter has been discontinued. The direct replacement for the Thermo Scientific G2 Fast Blotter is the Thermo Scientific Power Blotter (Cat. No. 22834). The 1-Step Transfer buffer (Cat. No. 84731) is still available.