DyLight™ 550 Maleimide
Thermo Scientific™

DyLight™ 550 Maleimide

Thermo Scientific DyLight 550 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 550 used to fluorescently label sulfhydryl-containing peptides,Read more
Have Questions?
Catalog NumberQuantity
622901 mg
Catalog number 62290
Price (USD)
461.00
Each
Quantity:
1 mg
Price (USD)
461.00
Each
Thermo Scientific DyLight 550 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 550 used to fluorescently label sulfhydryl-containing peptides, proteins and other biomolecular probes.

DyLight 550 provides vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives including Alexa Fluor™ 555, TRITC and Cy3™ dye for fluorescent applications over a broad pH range (pH 4-9). The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates.

Features of DyLight 550 Maleimide:

• High performance—DyLight 550 shows brighter fluorescence than Alexa Fluor 555, TRITC and Cy3 dye
• Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
• Efficient labeling methods—well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
• Optimized antibody labeling procedure—complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
• Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
• Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

DyLight 550 Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
DescriptionDyLight 550 Maleimide
Emission576
Excitation562
Label or DyeDyLight™ 550
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C. Shipped at ambient temperature.

Citations & References (1)

Citations & References
Abstract
Substrate degradation by the proteasome: a single-molecule kinetic analysis.
Authors:Lu Y, Lee BH, King RW, Finley D, Kirschner MW,
Journal:
PubMed ID:25859050
'To address how the configuration of conjugated ubiquitins determines the recognition of substrates by the proteasome, we analyzed the degradation kinetics of substrates with chemically defined ubiquitin configurations. Contrary to the view that a tetraubiquitin chain is the minimal signal for efficient degradation, we find that distributing the ubiquitins as ... More