Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD44 Antibodies

          Invitrogen

          CD44 Monoclonal Antibody (IM7), Super Bright™ 645, eBioscience™

          98 Published Figures
          129 References
          View all (269) CD44 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD44 Monoclonal Antibody (IM7), Super Bright™ 645, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          CD44 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD44 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD44 Antibody (64-0441-82) in Flow

          Staining of C57BL/6 splenocytes with Anti-Mouse CD3e FITC (Product # 11-0031-82) and 0.5 µg of Rat IgG2b kappa Isotype Control Super Bright 645 (Product # 64-4031-82) (left) or 0.5 µg of Anti-Human/Mouse CD44 Super Bright 645 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD44 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD44 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunocytochemistry (ICC/IF)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Immunohistochemistry (IHC)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)
          CD44 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          64-0441-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunocytochemistry (ICC/IF)

          -
          View 9 publications 9 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 108 publications 108 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), Super Bright™ 645, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          IM7

          Conjugate

          Super Bright™ 645 Super Bright™ 645 Super Bright™ 645
          • Unconjugated
          • Alexa Fluor 532 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 690
          • NovaFluor Blue 725
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 700
          • NovaFluor Red 710
          • NovaFluor Red 725
          • NovaFluor Red 755
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Yellow 570
          • NovaFluor Yellow 610
          • NovaFluor Yellow 660
          • NovaFluor Yellow 755
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          414/645 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2662590

          Product Specific Information

          Description: The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.

          Applications Reported: This IM7 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This IM7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          CD44 cell surface antigen is a 100 kDa type 1 transmembrane glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential. CD44 is expressed by hematopoietic, non-hematopoietic cells, epithelial tissues, and to filopodia in cultured keratinocytes. Further, bone marrow myeloid cells and memory T cells express CD44 at high levels, and peripheral B and T cells can upregulate the expression of CD44 in response to certain stimulatory events. Transcripts for the CD44 gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full-length nature of some of these variants have not been determined. Alternative splicing is the basis for the structural and functional diversity of the CD44 protein. Diseases associated with CD44 dysfunction include superficial keratitis and lichen sclerosus. CD44 also may be related to tumor metastasis formation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Cd44; CD44 molecule; CD44 molecule (Indian blood group); CD44 precursor; CD44R1; CDw44 antigen precursor; cell surface glycoprotein CD44; cell-cell and cell-matrix adhesion interaction glycoprotein isoform; cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; ECMR-III; ECMRIII; epican; extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; Hermes antigen; Hermes antigen gp90 homing receptor precursor; homing cell adhesion molecule; homing function and Indian blood group system; homing receptor; HUTCH-I; hyaluronan receptor; hyaluronan-binding protein; hyaluronate receptor; In(Lu) related-p80; Indian blood group antigen; lymphocyte antigen 24; PGP I; PGP-I; PGPI; phagocyte glycoprotein 1; phagocytic glycoprotein 1; phagocytic glycoprotein I; phosphoglycoprotein Pgp-1 (Ly24.2) precursor; sCD 44; sCD44; soluble CD 44; soluble CD44; unnamed protein product

          View more View less

          Gene Aliases: CDW44; CSPG8; ECM-III; ECMR-III; H-CAM; HCELL; HERMES; Hermes-1; HUTCH-1; HUTCH-I; LHR; Ly-24; MC56; MDU2; MDU3; MIC4; Pgp-1; Pgp1

          View more View less

          UniProt ID: (Human) P16070, (Mouse) P15379

          View more View less

          Entrez Gene ID: (Human) 960, (Mouse) 12505

          View more View less

          Function(s)
          transmembrane signaling receptor activity cytokine receptor activity protein binding collagen binding hyaluronic acid binding signaling receptor activity cargo receptor activity type II transforming growth factor beta receptor binding epidermal growth factor receptor binding channel regulator activity protein kinase binding cytokine binding phosphoprotein binding
          Process(es)
          regulation of cell growth branching involved in ureteric bud morphogenesis wound healing involved in inflammatory response positive regulation of adaptive immune response negative regulation of mature B cell apoptotic process receptor-mediated endocytosis inflammatory response cell adhesion anatomical structure morphogenesis positive regulation of gene expression Wnt signaling pathway cell migration cytokine-mediated signaling pathway hyaluronan catabolic process neuron projection development positive regulation of neutrophil apoptotic process positive regulation of heterotypic cell-cell adhesion macrophage fusion T cell activation negative regulation of apoptotic process negative regulation of DNA damage response, signal transduction by p53 class mediator wound healing, spreading of cells cellular response to fibroblast growth factor stimulus negative regulation of regulatory T cell differentiation system development negative regulation of inflammatory response NK T cell activation branching involved in prostate gland morphogenesis positive regulation of ERK1 and ERK2 cascade monocyte aggregation regulation of modification of postsynaptic structure postsynapse organization positive regulation of monocyte aggregation negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator regulation of leukocyte apoptotic process regulation of lamellipodium morphogenesis negative regulation of CD4-positive, alpha-beta T cell proliferation endocytosis cell-matrix adhesion regulation of apoptotic process cartilage development cell-cell adhesion carbohydrate derivative transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch