Inquire about OEM or Commercial Supply version of this product here.
Dynabeads™ MyOne™ Tosylactivated
Invitrogen™

Dynabeads™ MyOne™ Tosylactivated

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
6550210 mL
655012 mL
Catalog number 65502
Price (USD)
4,830.00
Each
Quantity:
10 mL
Price (USD)
4,830.00
Each

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated. Optimal antibody orientation makes these beads an excellent choice for the immunoprecipitation of proteins and protein complexes.

Ideal for Immunoprecipitation of protein complexes:
Low background and covalent linking of antibodies to the bead surface make Dynabeads Tosylactivated an excellent choice for Immunoprecipitation of proteins and protein complexes (Co-Immunoprecipitation, Co-IP). Gentle and rapid magnetic concentration of the beads and short incubation times (possible due to fast surface-based binding kinetics) make Dynabeads Tosylactivated an excellent choice for the immunoprecipitation of highly-labile or and/or transient (short-lived) protein complexes.

Bead Surface Characteristics for Dynabeads Tosylactivated:

  • p-toluene-sulfonyl (Tosyl) groups
  • Hydrophobic, pH Neutral
  • Covalent binding by primary amine (NH2) or sulphydryl (SH) groups

Ultra rapid protocols:

  • Identify members of protein complexes in minutes rather than hours
  • Temporal resolution short enough to identify transient and labile complexes
  • Identify binding partners that cannot be identified with longer protocols
  • Rapid protocols further reduce the already ultra-low background binding
  • Stronger signal to noise-ratios

Applications:

  • IP of proteins and protein complexes
  • Couple functional enzymes to bead surface for downstream assays
  • Couple peptides to bead surface to identify binders

Purify peptides, proteins and enzymes that are:

  • Transiently stable
  • Structurally intact
  • Temperature labile
  • In their native conformation and functional

Coupling procedure outline:
Covalent coupling is performed overnight by incubating the desired ligand with the Dynabeads Tosylactivated. Ligands commonly coupled to Dynabeads Tosylactivated include peptides and proteins (e.g. antibodies for Immunoprecipitation or Co-Immunoprecipitation). Coupling occurs at neutral to high pH and at 37°C. We recommend coupling at pH 8.5-9.5, but for pH labile ligands, coupling can be performed in an alternative buffer at pH 7.4.

Upon completion of the ligand coupling step, the actual Dynabeads Tosylactivated surface coating will be rendered inert, resulting in low non-specific binding.

Binding capacity per milligram beads:
Varies depending on ligand, (e.g. 14-19 μg IgG)

About Dynabeads:
Dynabeads are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling ligands coupled to the beads to continuously interact with the entire sample volume. The superparamagnetic beads are pulled to the tube walls by transferring the tube to a rack containing a strong magnetic field. Strong magnetic fields quickly pull the beads to the tube wall, allowing for easy and complete removal of the supernatant by pipette. Washing steps are performed similarly.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration100 mg/mL, 100 mg/mL
Elution Volume Min5 μL
FormatBeads in solution
High-throughput CompatibilityHigh-throughput Compatible
Purity or Quality GradeResearch Grade
Quantity10 mL
Sample TypeAny Sample Type
Shipping ConditionRoom Temperature
Surface Functionalityp-Toluene-Sulfonyl, Tosylactivated
Product LineDYNAL, Dynabeads, MyOne
TypeTosylactivated Bead
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.

Frequently asked questions (FAQs)

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.