Nalgene™ Large Polypropylene Sterilizing Pans, 14.2L - FAQs
Nalgene™ Large Polypropylene Sterilizing Pans, 14.2L - FAQs
View additional product information for Nalgene™ Large Polypropylene Sterilizing Pans - FAQs (6900-0020PK, 6900-0010PK, 6900-0020, 6900-0010)
1 product FAQs found
Do you offer any trays or pans for secondary containment of reagent bottles?
Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.