SureSTART™ Polypropylene Snap Top HPLC and PFAS Vials with Fixed-Insert, Level 1, 0.30 mL, Conical Insert, 5000/Pack - FAQs
SureSTART™ Polypropylene Snap Top HPLC and PFAS Vials with Fixed-Insert, Level 1, 0.30 mL, Conical Insert, 5000/Pack - FAQs
View additional product information for SureSTART™ Polypropylene Snap Top HPLC and PFAS Vials with Fixed-Insert, Level 1 - FAQs (6ERV11-08PPFBF, 6ERV11-08PPCT, 6ERV11-03PPC, 6ERV11-03PPCT, 6ERV11-08PPC, 6ERV11-08PPCF, 6ERV11-03PPCF, 6ERV11-08PPFB, 6ERV11-08PPFBT)
2 product FAQs found
What are SureSTART fixed insert vials?
During manufacture, the outer vial has a glass micro-insert precisely positioned inside and this is permanently fused to the base of the outer vial. This gives a 12 mm x 32 mm microvial with a capacity of 350 µL with a narrow reservoir with <10 µL capacity. A fixed insert vial removes the need to assemble a vial and insert prior to use.
What are SureSTART microvials?
Microvials are suitable for use when sample volume is limited or when you need to get maximum recovery from the vial. These vials allow small volumes to be sampled accurately by autosamplers with a recoverable volume of much less than 100 µL. High recovery, total recovery, and vials with fixed inserts are all classed as microvials.