SureSTART™ 10 mL Glass Crimp Top Headspace Vials, Level 3, Clear, 9.5 mL, High Recovery - FAQs
SureSTART™ 10 mL Glass Crimp Top Headspace Vials, Level 3, Clear, 9.5 mL, High Recovery - FAQs
View additional product information for SureSTART™ 10 mL Glass Crimp Top Headspace Vials, Level 3 - FAQs (6PCV10-1FT, 6PCV10-V1, 6PCV10-1F)
1 product FAQs found
What are SureSTART High Recovery vials?
These one-piece microvials have an internal conical reservoir which collects liquid in a 30 µL volume for extraction with a conventional autosampler from a total usable volume of 1.7 mL. The outer dimension for these vials is 12 x 32 mm.