SureSTART™ 0.3 mL Clear Glass Inserts (Level 3) for 2 mL Vials, Conical with PE Polyspring, 5000/pack - FAQs
SureSTART™ 0.3 mL Clear Glass Inserts (Level 3) for 2 mL Vials, Conical with PE Polyspring, 5000/pack - FAQs
View additional product information for SureSTART™ Glass Polyspring Inserts for 2 mL HPLC & GC Vials, Level 3 - FAQs (6PME05FS1, 6PME03C1SPT, 6PME03C1SSPF, 6PME04C1F, 6PME04CPR1, 6PME03C1SPG, 6PME03C1SPF, 6PME04C1T, 6PME03C1SSPT, 6PME03C1SSP, 6PME04C1, 6PME03C1SP)
1 product FAQs found
What are SureSTART micro inserts?
The micro inserts have a small total volume capacity of <400 µL and can be classified as either Polyspring, conical, or flat-bottom within the SureSTART Collection. Micro inserts are placed and sealed inside a vial so they can be used in conventional autosampler racks. The smaller 6 mm diameter of the insert gives better sample recovery than a routine flat based vial with conical inserts being most effective with residual volumes of 1-2 µL. Many are held in place by a Polyspring, which cushions and centers the insert.