SureSTART™ 1.5 mL Total Recovery Glass Snap Top (U)HPLC and GC Vials, Level 3, Clear, Total Recovery, 1000/Pack - FAQs
SureSTART™ 1.5 mL Total Recovery Glass Snap Top (U)HPLC and GC Vials, Level 3, Clear, Total Recovery, 1000/Pack - FAQs
View additional product information for SureSTART™ 1.5 mL Total Recovery Glass Snap Top (U)HPLC and GC Vials, Level 3 - FAQs (6PRV11-TR1T, 6PRV11-TR1)
2 product FAQs found
What are SureSTART Total Recovery vials?
These one-piece microvials have a very low residual volume due to a specially shaped deep and narrow inner reservoir which collects the liquid in the base so compatible autosamplers can collect all but 1-2 microliters of sample while having a total sample volume of 1.2 mL. The outer dimension for these vials is 12 x 32 mm.
What are SureSTART microvials?
Microvials are suitable for use when sample volume is limited or when you need to get maximum recovery from the vial. These vials allow small volumes to be sampled accurately by autosamplers with a recoverable volume of much less than 100 µL. High recovery, total recovery, and vials with fixed inserts are all classed as microvials.