Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MARVELD2 Antibodies

          Invitrogen

          MARVELD2 Recombinant Rabbit Monoclonal Antibody (54H19L38)

          Advanced Verification
          18 Published Figures
          32 References
          View all (7) MARVELD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MARVELD2 Recombinant Rabbit Monoclonal Antibody (54H19L38)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (18)
          • Advanced Verification (2)
          MARVELD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MARVELD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MARVELD2 Antibody (700191) in ICC/IF

          Immunofluorescence analysis of Tricellulin was done on 90% confluent log phase Caco-2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with Tricellulin Rabbit Monoclonal Antibody (Product # 700191) at 1 µg/mL in 1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MARVELD2 Antibody in Immunocytochemistry (ICC/IF)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Immunoprecipitation (IP)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Immunocytochemistry, Proximity Ligation Assay (PLA) (ICC/IF, PLA)
          MARVELD2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          MARVELD2 Antibody in Immunocytochemistry (ICC/IF)
          MARVELD2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          MARVELD2 Antibody in Immunohistochemistry (IHC)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody in Western Blot (WB)
          MARVELD2 Antibody
          MARVELD2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MARVELD2 Recombinant Rabbit Monoclonal Antibody (54H19L38)

          Product Details

          700191

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-3 µg/mL
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          View 8 publications 8 publications

          Immunoprecipitation (IP)

          5 µg
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Dog, Human, Mouse, Pig

          Host/Isotype

          Rabbit / IgG

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          54H19L38

          Immunogen

          A recombinant protein corresponding to amino acids 369-558 of Q8N4S9.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2-7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Wet ice

          RRID

          AB_2532298

          Product Specific Information

          This antibody is predicted to react with chimpanzee, mouse, rat, bovine and Xenopus based on sequence homology.

          Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain.

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          Tight junctions form an important barrier of paracellular transport in epithelial cells. Sealing of two adjacent cells at bicellular tight junctions, a point where three adjacent cells are in contact with each other. Tricellulin is the first protein identified that specifically concentrates in tricellular tight junctions. This protein has four membrane spanning domains, similarly to claudins. Tricellulin expression is high in epithelium-derived tissues, such as small intestine, kidney and lung. Functional evidence for the role of tricellulin in tight junction formation comes from siRNA studies, where suppression of its expression leads to compromised epithelial barrier and tight junction formation. Loss of function in tricellulin mutants missing all or most of a conserved region in the cytosolic domain which binds to the cytosolic scaffolding protein ZO-1, indicate that interaction with other known tight junction proteins plays an important role for the function of tricellulin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MARVEL (membrane-associating) domain containing 2; TRIC; TRICbeta; tricellular junction protein; tricellulin; unnamed protein product

          View more View less

          Gene Aliases: DFNB49; MARVD2; MRVLDC2; Tric

          View more View less

          UniProt ID: (Human) Q8N4S9

          View more View less

          Entrez Gene ID: (Human) 153562

          View more View less

          Function(s)
          protein binding
          Process(es)
          sensory perception of sound cell-cell junction organization establishment of endothelial barrier bicellular tight junction assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch