Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RUNX2 Antibodies
          This clone is available in multiple conjugated formats in both full and sample sizes. For more options, search using the clone ID "6H4L27" or explore the "Additional formats" section on this page.

          Invitrogen

          RUNX2 Recombinant Rabbit Monoclonal Antibody (6H4L27), Alexa Fluor™ Plus 555

          Advanced Verification
          View all (51) RUNX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RUNX2 Recombinant Rabbit Monoclonal Antibody (6H4L27), Alexa Fluor™ Plus 555

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Advanced Verification (2)
          RUNX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RUNX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RUNX2 Antibody (702489RP555) in WB

          Western blot analysis was performed on Whole cell extracts (40 µg lysate) of A-431 (Lane 1), SH-SY5Y (Lane 2), HeLa (Lane 3), SW480 (Lane 4), LNCaP (Lane 5), Raji (Lane 6), Hep G2 (Lane 7) and MCF7 (Lane 8) and blocking with Blocker™ FL Fluorescent Blocking Buffer (Product # 37565). The blots were probed with RUNX2 Recombinant Rabbit Monoclonal Antibody (6H4L27), Alexa Fluor™ Plus 555 (Product # 702489RP555, 1:200). A ~56 kDa band corresponding to RUNX2 was observed in all cell lysates tested. Known quantity of protein samples were electro... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RUNX2 Antibody in Western Blot (WB)
          RUNX2 Antibody in Western Blot (WB)
          RUNX2 Antibody in Flow Cytometry (Flow)
          RUNX2 Antibody
          RUNX2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          702489RP555

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Flow Cytometry (Flow)

          1 µg/test
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          6H4L27

          Immunogen

          Protein corresponding to human RUNX2 [aa235-aa400]

          Conjugate

          Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555
          • Unconjugated
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 647 
          • Request custom conjugation

          Excitation/Emission Max

          558/572 nm View spectra spectra

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          proprietary buffer, pH 6.8

          Contains

          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_3679122

          Product Specific Information

          Alexa Fluor™ Plus recombinant rabbit antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro.

          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.

          Applications Tested: This 6H4L27 antibody has been tested by western blot and flow cytometric analysis of U-2 OS cells. This may be used for western blot at 5 µg/mL and for flow cytometry at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 553 nm; Emission: 568 nm; Laser: Yellow Laser

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: acute myeloid leukemia 3 protein; AML-3; core-binding factor, runt domain, alpha subunit 1; oncogene AML-3; osteoblast-specific transcription factor 2; PEA2-alpha A; PEBP2-alpha A; polyomavirus enhancer-binding protein 2 alpha A subunit; runt related transcription factor 2; RUNX 2; RUNX-2; SL3-3 enhancer factor 1 alpha A subunit; SL3/AKV core-binding factor alpha A subunit

          View more View less

          Gene Aliases: AML3; CBF-alpha-1; CBFA1; CCD; CCD1; CLCD; OSF-2; OSF2; PEA2aA; PEBP2aA

          View more View less

          UniProt ID: (Human) Q13950

          View more View less

          Entrez Gene ID: (Human) 860

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding ATP binding sequence-specific double-stranded DNA binding
          Process(es)
          ossification osteoblast differentiation chondrocyte differentiation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transmembrane receptor protein serine/threonine kinase signaling pathway hemopoiesis neuron differentiation BMP signaling pathway regulation of cell differentiation positive regulation of osteoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter SMAD protein signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0