Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Parkin Antibodies

          Invitrogen

          Parkin Recombinant Rabbit Monoclonal Antibody (21H24L9), HRP

          Advanced Verification
          View all (42) Parkin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Parkin Recombinant Rabbit Monoclonal Antibody (21H24L9), HRP

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          Parkin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Parkin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Parkin Antibody (702785RHRP) in WB

          Western blot analysis was performed on Whole cell extracts (60 µg lysate) of PC-12 (Lane 1), tissue extracts of Mouse Brain (Lane 2), Mouse Lung (Lane 3), Rat Lung (Lane 4) and Rat Brain (Lane 5). The blots were probed with Parkin Recombinant Rabbit Monoclonal Antibody, HRP (Product # 702785RHRP, 1:1,000). A 50 kDa band corresponding to Parkin was observed in PC-12 cell line and tissues tested. Known quantity of protein samples were electrophoresed using Novex®NuPAGE®4-12% Bis-Tris gel (Product # NP0322BOX), XCell SureLock™ Electrophoresis... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Parkin Antibody in Western Blot (WB)
          Parkin Antibody in Western Blot (WB)
          Parkin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Parkin Recombinant Rabbit Monoclonal Antibody (21H24L9), HRP

          Product Details

          702785RHRP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          21H24L9

          Immunogen

          Protein corresponding to human Parkin (aa1-aa465)
          3D Epitope / Immunogen

          Conjugate

          HRP HRP HRP
          • Unconjugated
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2-7.4, with proprietary stabilizer

          Contains

          proprietary preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2942251

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, lot-to-lot consistency, and broader immunoreactivity to diverse targets due to a larger rabbit immune repertoire.

          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Target Information

          The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E3 ubiquitin ligase; E3 ubiquitin protein ligase; parkin; E3 ubiquitin protein ligase; parkinson protein 2; OTTHUMP00000017562; OTTHUMP00000017563; OTTHUMP00000017564; parkin 2; Parkin isoform 1; parkin protein; parkin RBR E3 ubiquitin protein ligase 2; parkin variant SV5DEL; Parkinson disease (autosomal recessive, juvenile) 2, parkin; parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2 E3 ubiquitin protein ligase (parkin) isoform PARK2_1; parkinson protein 2 E3 ubiquitin protein ligase (parkin) isoform PARK2_2; parkinson protein 2 E3 ubiquitin protein ligase (parkin) isoform PARK2_3; parkinson protein 2, E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin); parkinson protein 2; E3 ubiquitin protein ligase; ubiquitin ligase E3; ubiquitin ligase E3; contains additional exon 1b between canonical exons 1 and 2; ubiquitin ligase E3; lacks exon 5; ubiquitin ligase E3; lacks exon 9; unnamed protein product

          View more View less

          Gene Aliases: AR-JP; LPRS2; Park; PARK2; PDJ

          View more View less

          UniProt ID: (Human) O60260, (Rat) Q9JK66, (Mouse) Q9WVS6

          View more View less

          Entrez Gene ID: (Human) 5071, (Rat) 56816, (Mouse) 50873

          View more View less

          Function(s)
          G-protein coupled receptor binding transcription corepressor activity actin binding ubiquitin-protein transferase activity protein binding beta-catenin binding zinc ion binding tubulin binding SH3 domain binding enzyme binding kinase binding protein kinase binding PDZ domain binding Hsp70 protein binding heat shock protein binding ubiquitin conjugating enzyme binding ubiquitin protein ligase binding identical protein binding histone deacetylase binding ubiquitin binding phospholipase binding macromolecular complex binding chaperone binding ubiquitin protein ligase activity cullin family protein binding ubiquitin-specific protease binding F-box domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter protein polyubiquitination mitochondrial fission mitophagy mitophagy ubiquitin-dependent protein catabolic process protein monoubiquitination response to oxidative stress response to unfolded protein mitochondrion organization central nervous system development adult locomotory behavior response to xenobiotic stimulus positive regulation of catabolic process proteasomal protein catabolic process regulation of autophagy positive regulation of gene expression negative regulation of gene expression positive regulation of mitochondrial fusion negative regulation of mitochondrial fusion regulation of mitochondrion organization regulation of glucose metabolic process positive regulation of mitochondrial membrane potential free ubiquitin chain polymerization regulation of dopamine secretion response to muscle activity macroautophagy protein ubiquitination protein deubiquitination regulation of anatomical structure morphogenesis protein catabolic process regulation of protein ubiquitination regulation of protein stability protein destabilization negative regulation of actin filament bundle assembly regulation of lipid transport positive regulation of proteasomal ubiquitin-dependent protein catabolic process negative regulation of glucokinase activity cellular response to oxidative stress cellular response to unfolded protein response to endoplasmic reticulum stress protein K29-linked ubiquitination positive regulation of insulin secretion involved in cellular response to glucose stimulus ERAD pathway regulation of dopamine metabolic process dopamine metabolic process positive regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of neuron apoptotic process mitochondrial fragmentation involved in apoptotic process protein K27-linked ubiquitination negative regulation by host of viral genome replication positive regulation of protein catabolic process positive regulation of transcription from RNA polymerase II promoter negative regulation of JNK cascade negative regulation of insulin secretion modulation of synaptic transmission protein stabilization response to corticosterone positive regulation of neurotransmitter uptake mitochondrion localization protein autoubiquitination regulation of mitochondrial membrane potential negative regulation of synaptic transmission, glutamatergic regulation of necroptotic process regulation of canonical Wnt signaling pathway parkin-mediated mitophagy in response to mitochondrial depolarization neuron cellular homeostasis protein K63-linked ubiquitination protein localization to mitochondrion aggresome assembly protein K48-linked ubiquitination protein K11-linked ubiquitination cellular response to manganese ion protein K6-linked ubiquitination negative regulation of canonical Wnt signaling pathway positive regulation of mitochondrial fission negative regulation of release of cytochrome c from mitochondria negative regulation of mitochondrial fission negative regulation of excitatory postsynaptic potential cellular response to toxic substance regulation of postsynaptic membrane neurotransmitter receptor levels mitochondrion to lysosome transport regulation protein catabolic process at presynapse regulation of synaptic vesicle endocytosis regulation of cellular response to oxidative stress positive regulation of mitophagy positive regulation of proteasomal protein catabolic process negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator positive regulation of protein linear polyubiquitination regulation of synaptic vesicle transport positive regulation of tumor necrosis factor-mediated signaling pathway cellular response to dopamine negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of reactive oxygen species biosynthetic process regulation of secretion by cell negative regulation of exosomal secretion regulation of protein localization to mitochondrion positive regulation of dendrite extension negative regulation of spontaneous neurotransmitter secretion response to curcumin cellular response to L-glutamine cellular response to hydrogen sulfide positive regulation of type 2 mitophagy cellular response to L-glutamate positive regulation of retrograde transport, endosome to Golgi negative regulation of intralumenal vesicle formation positive regulation of protein localization to membrane amyloid fibril formation regulation of reactive oxygen species metabolic process negative regulation of reactive oxygen species metabolic process positive regulation of ATP biosynthetic process synaptic transmission, dopaminergic startle response learning locomotory behavior synaptic transmission, glutamatergic norepinephrine metabolic process regulation of neurotransmitter secretion dopamine uptake involved in synaptic transmission
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0