I am interested in RNAse-free 0.1M DTT and I am considering USB Dithiothreitol (DTT), 0.1M Solution (Cat. No. 707265ML). What does USB stands for and is this the regular DTT?
Yes, this is the regular DTT. "USB" is a former brand name.
Search
View additional product information for USB Dithiothreitol (DTT), 0.1M Solution - FAQs (707265ML)
1 product FAQs found