Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Connexin 26 Antibodies

          Invitrogen

          Connexin 26 Polyclonal Antibody

          Advanced Verification
          43 Published Figures
          101 References
          View all (11) Connexin 26 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Connexin 26 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (43)
          • Advanced Verification (1)
          Connexin 26 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Connexin 26 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 47

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 26 Antibody (71-0500) in IHC (P)

          Immunohistochemistry analysis of Connexin 26 showing staining in the membrane of paraffin-embedded mouse liver tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at room temperature... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 26 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 26 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (IHC)
          Connexin 26 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Connexin 26 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody in Western Blot (WB)
          Connexin 26 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          71-0500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 38 publications 38 publications

          Immunohistochemistry (IHC)

          -
          View 23 publications 23 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 6 publications 6 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 18 publications 18 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Chicken, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide corresponding to a portion of the cytoplasmic loop of rat connexin 26.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533971

          Product Specific Information

          71-0500 was used in the immunofluorescence and western blot analysis to successfully detect Connexin26 in mouse liver sections and homogenate, respectively.

          Target Information

          Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: connexin 26; connexin e; connexin-26; gap junction channel protein connexin 26; gap junction membrane channel protein beta 2; unnamed protein product

          View more View less

          Gene Aliases: Cnx26; Cx26; CXN-26; Cxne; Gjb-2

          View more View less

          UniProt ID: (Rat) P21994, (Mouse) Q00977

          View more View less

          Entrez Gene ID: (Rat) 394266, (Mouse) 14619

          View more View less

          Function(s)
          gap junction channel activity calcium ion binding protein binding identical protein binding gap junction channel activity involved in cell communication by electrical coupling
          Process(es)
          response to ischemia cell communication cell-cell signaling female pregnancy cell communication by electrical coupling gap junction assembly response to estradiol response to lipopolysaccharide response to retinoic acid response to progesterone cellular response to oxidative stress response to human chorionic gonadotropin decidualization inner ear development transmembrane transport cellular response to glucagon stimulus cellular response to dexamethasone stimulus epididymis development gap junction-mediated intercellular transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch