Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Connexin 30 Antibodies

          Invitrogen

          Connexin 30 Polyclonal Antibody (Z-PP9)

          87 Published Figures
          138 References
          View all (4) Connexin 30 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Connexin 30 Polyclonal Antibody (Z-PP9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (87)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 90

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 30 Antibody (71-2200) in ICC/IF

          Immunofluorescence analysis of Connexin-30 was done on 70% confluent log phase Caco-2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with Connexin-30 Rabbit polyclonal Antibody (Product # 71-2200) at 2 µg/mL in 1% BSA and incubated for 3 hours at room te... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 30 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Connexin 30 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 30 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 30 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 30 Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Immunohistochemistry (IHC)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Western Blot (WB)
          Connexin 30 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Connexin 30 Polyclonal Antibody (Z-PP9)

          Product Details

          71-2200

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 38 publications 38 publications

          Immunohistochemistry (IHC)

          -
          View 48 publications 48 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 13 publications 13 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          View 25 publications 25 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 9 publications 9 publications
          Product Specifications

          Species Reactivity

          Cat, Human, Mouse, Rat

          Published species

          Cat, Chicken, Guinea pig, Human, Mouse, Non-human primate, Rat, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Clone

          Z-PP9

          Immunogen

          A synthetic peptide derived from the unique C-terminus of the mouse Connexin 30 protein
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533979

          Product Specific Information

          This antibody is specific for the Connexin 30. Cross-reactivity with the highly related Connexin 26 protein or with other Connexin family members has not been detected. Reactivity of this antibody has been confirmed by western blotting using cell lysates derived from adult rat, mouse, human, and cat brains.

          Target Information

          Gap junctions allow the transport of ions and metabolites between the cytoplasm of adjacent cells. They are formed by two hemichannels, made up of six connexin proteins assembled in groups. Each connexin protein has four transmembrane segments, two extracellular loops, a cytoplasmic loop formed between the two inner transmembrane segments, and the N- and C-terminus both being in the cytoplasm. The specificity of the gap junction is determined by which connexin proteins comprise the hemichannel. In the past, connexin protein names were based on their molecular weight, however the new nomenclature uses sequential numbers based on which form of the gap junction is present. This gene encodes one of the connexin proteins. Mutations in this gene have been found in some forms of deafness and in some families with hidrotic ectodermal dysplasia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: connexin 30; connexin f; connexin-30; Cx30; ectodermal dysplasia 2, hidrotic (Clouston syndrome); gap junction membrane channel protein beta 6; gap junction protein beta 6; gap junction protein, beta 6 (connexin 30); gap junction protein, beta 6, 30kDa; unnamed protein product

          View more View less

          Gene Aliases: CX30; Cxnf; D14Bwg0506e; DFNA3; DFNA3B; DFNB1B; ECTD2; ED2; EDH; HED; HED2

          View more View less

          UniProt ID: (Human) O95452, (Mouse) P70689

          View more View less

          Entrez Gene ID: (Human) 10804, (Rat) 84403, (Mouse) 14623

          View more View less

          Function(s)
          gap junction channel activity protein binding microtubule binding beta-tubulin binding actin filament binding gap junction channel activity involved in cell communication by electrical coupling
          Process(es)
          sinoatrial node development cell communication cell-cell signaling sensory perception of sound cell communication by electrical coupling gap junction assembly maintenance of blood-brain barrier transmembrane transport gap junction-mediated intercellular transport negative regulation of cell proliferation response to lipopolysaccharide ear morphogenesis inner ear development response to electrical stimulus cellular response to glucose stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch