Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAB11A Antibodies

          Invitrogen

          RAB11A Polyclonal Antibody

          Advanced Verification
          49 Published Figures
          152 References
          View all (23) RAB11A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RAB11A Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (12)
          • Published Figures (49)
          • Advanced Verification (1)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 62

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB11A Antibody (71-5300) in ICC/IF

          Immunofluorescence analysis of Ras-related protein Rab-11A was performed using 70% confluent log phase Hep G2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with RAB11A Polyclonal Antibody (Product # 71-5300) at 1:100 in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB11A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB11A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry, Dot Blot (ICC/IF, DB)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunocytochemistry (ICC/IF)
          RAB11A Antibody in Immunohistochemistry (Frozen) (IHC (F))
          RAB11A Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          RAB11A Antibody in Immunohistochemistry (IHC)
          RAB11A Antibody in Immunohistochemistry (IHC)
          RAB11A Antibody in Immunohistochemistry, Dot Blot (IHC, DB)
          RAB11A Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          RAB11A Antibody in Immunohistochemistry (IHC)
          RAB11A Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          RAB11A Antibody in Immunohistochemistry (IHC)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RAB11A Antibody in Western Blot (WB)
          RAB11A Antibody in Immunoprecipitation (IP)
          RAB11A Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RAB11A Polyclonal Antibody

          Product Details

          71-5300

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 44 publications 44 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1-5 µg
          View 82 publications 82 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          -

          Immunoprecipitation (IP)

          5 µg
          View 3 publications 3 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          Assay-dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate, Rabbit, Rat

          Published species

          Bacteria, Dog, Fruit fly, Hamster, Human, Mouse, Non-human primate, Rat, Rhesus monkey

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the C-terminal hypervariable region of the human Rab11a sequence. This peptide shares 10 of 17 residues with the related Rab11b protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533987

          Product Specific Information

          This polyclonal antibody reacts specifically with the human Rab11A protein. Cross-reactivity with other Rab family members has not been detected.

          Positive control: Human HEK-293 cells

          Target Information

          Rab11A is a member of the YPT-1 subfamily of Ras-related GTP-binding proteins. Rab11A mRNA has been detected in a variety of tissues including brain, testis, spleen, heart, and gastrointestinal mucosa. In the gastric fundus the Rab11A protein is expressed in parietal cell tubulovesicles membranes. Expression of the Rab11A protein appears to occur in discrete epithelial cell populations where it localizes to apical vesicular populations. Recent studies have shown Rab11A is essential for resecretion of alpha-synuclein out of neuronal cells via endosome recycling and is required for exocytosis of discoidal/fusiform vesicles of bladder umbrella cells. Rab11A is also implicated in biogenesis of Birbeck granules and endosomal activation of the PI3K/AKT signaling pathway via G beta gamma trafficking.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 24KG; GTPase; RAB 11A, member oncogene family; rab-11; rab11; rab11 GTP-binding protein; RAB11A; tubulovesicle-associated protein; unnamed protein product

          View more View less

          Gene Aliases: RAB11; YL8

          View more View less

          UniProt ID: (Dog) P62490, (Rabbit) P62493, (Human) P62491, (Mouse) P62492, (Rat) P62494

          View more View less

          Entrez Gene ID: (Dog) 403957, (Rabbit) 100009072, (Human) 8766, (Mouse) 53869, (Rat) 81830

          View more View less

          Function(s)
          GTPase activity GTP binding microtubule binding protein domain specific binding myosin V binding obsolete small monomeric GTPase activity protein binding dynein light intermediate chain binding
          Process(es)
          exocytosis microtubule-based movement mitotic metaphase plate congression positive regulation of epithelial cell migration positive regulation of G2/M transition of mitotic cell cycle vesicle-mediated transport astral microtubule organization neuron projection development melanosome transport endocytic recycling regulation of cytokinesis protein localization to cell surface multivesicular body assembly Golgi to plasma membrane protein transport positive regulation of cilium assembly regulation of long-term neuronal synaptic plasticity regulation of protein transport establishment of vesicle localization regulation of vesicle-mediated transport protein localization to cilium protein transmembrane transport establishment of protein localization to organelle protein localization to plasma membrane establishment of protein localization to membrane mitotic spindle assembly neurotransmitter receptor transport, endosome to postsynaptic membrane vesicle-mediated transport in synapse synaptic vesicle endosomal processing amyloid-beta clearance by transcytosis regulation of cilium assembly positive regulation of protein localization to plasma membrane positive regulation of mitotic cytokinetic process regulation of protein localization to centrosome protein localization to cleavage furrow exosomal secretion regulation of endocytic recycling regulation of multivesicular body size positive regulation of axon extension plasma membrane to endosome transport vesicle-mediated transport to the plasma membrane intracellular cholesterol transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0