Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • STAT5 alpha Antibodies

          Invitrogen

          Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody

          Advanced Verification
          33 Published Figures
          50 References
          View all (62) STAT5 alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (33)
          • Advanced Verification (2)
          Phospho-STAT5 alpha (Tyr694) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-STAT5 alpha (Tyr694) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 37

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-STAT5 alpha (Tyr694) Antibody (71-6900) in ICC/IF

          Immunofluorescent analysis of STAT5 (pY694) was done on 70% confluent log phase HeLa cells; untreated & treated (serum starved for 16 hours followed by treatment with 20 ng/mL IFN alpha for 1 hour). The cells were fixed with 4% paraformaldehyde for 15 minutes; permeabilized with 0.25% Triton™ X-100 for 10 minutes followed by blocking with 5% BSA for 1 hour at room temperature. The cells were incubated with ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-STAT5 alpha (Tyr694) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Immunohistochemistry (IHC)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot, ChIP Assay (WB, ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in Western Blot (WB)
          Phospho-STAT5 alpha (Tyr694) Antibody in ChIP Assay (ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody in ChIP Assay (ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody in ChIP Assay (ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody in ChIP Assay (ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody in ChIP Assay (ChIP)
          Phospho-STAT5 alpha (Tyr694) Antibody
          Phospho-STAT5 alpha (Tyr694) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody

          Product Details

          71-6900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 34 publications 34 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 4 publications 4 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications

          Gel Shift (EMSA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Sheep, Rat

          Published species

          Hamster, Human, Mouse, Non-human primate, Rat, Tag, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic tyrosine-phosphorylated peptide encompassing the conserved C-terminal tyrosine phosphorylation site (Y694) of murine STAT5. This peptide is conserved in human, bovine, sheep, and rat STAT5 proteins
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533991

          Target Information

          STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis secretory sperm binding protein; lacks exon 18; mammary gland factor; mammary gland factor STAT5A; signal transducer and activator of transcription 5; signal transducer and activator of transcription factor 5A; STAT; STAT5A, Mammary Gland Factor; STAT5B; transcription factor; unnamed protein product

          View more View less

          Gene Aliases: MGF; STAT5; STAT5A/MGF; STATA5

          View more View less

          UniProt ID: (Sheep) P42231, (Human) P42229, (Bovine) Q95115, (Rat) Q62771, (Mouse) P42230

          View more View less

          Entrez Gene ID: (Sheep) 443419, (Human) 6776, (Bovine) 282375, (Rat) 24918, (Mouse) 20850

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding phosphate ion binding sequence-specific DNA binding DNA-binding transcription factor binding promoter-specific chromatin binding double-stranded DNA binding DNA-binding transcription factor
          Process(es)
          positive regulation of endothelial cell proliferation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter defense response signal transduction JAK-STAT cascade cytokine-mediated signaling pathway taurine metabolic process eosinophil differentiation interleukin-15-mediated signaling pathway interleukin-4-mediated signaling pathway reelin-mediated signaling pathway interleukin-5-mediated signaling pathway interleukin-2-mediated signaling pathway interleukin-7-mediated signaling pathway interleukin-9-mediated signaling pathway interleukin-3-mediated signaling pathway thrombopoietin-mediated signaling pathway regulation of multicellular organism growth regulation of cell proliferation response to peptide hormone positive regulation of blood vessel endothelial cell migration positive regulation of transcription from RNA polymerase II promoter JAK-STAT cascade involved in growth hormone signaling pathway regulation of hemopoiesis negative regulation of T-helper 17 cell lineage commitment luteinization natural killer cell differentiation female pregnancy lactation positive regulation of cell proliferation regulation of steroid metabolic process lipid storage regulation of cell adhesion regulation of epithelial cell differentiation mammary gland development positive regulation of interleukin-2 production positive regulation of multicellular organism growth positive regulation of activated T cell proliferation T cell homeostasis negative regulation of apoptotic process positive regulation of B cell differentiation negative regulation of erythrocyte differentiation positive regulation of mitotic cell cycle development of secondary female sexual characteristics positive regulation of inflammatory response transcription from RNA polymerase II promoter gene expression peptidyl-tyrosine phosphorylation response to nutrient levels positive regulation of natural killer cell differentiation negative regulation of mast cell apoptotic process T cell differentiation in thymus response to ethanol positive regulation of gamma-delta T cell differentiation positive regulation of lymphocyte differentiation positive regulation of natural killer cell mediated cytotoxicity development of secondary male sexual characteristics Peyer's patch development gland development mammary gland involution positive regulation of mast cell differentiation prostate gland epithelium morphogenesis epithelial cell differentiation involved in prostate gland development mammary gland epithelium development positive regulation of mast cell proliferation cellular response to cytokine stimulus cellular response to granulocyte macrophage colony-stimulating factor stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-x4rxd:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0