Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 1 Antibodies

          Invitrogen

          Claudin 1 Polyclonal Antibody (MH25)

          Advanced Verification
          88 Published Figures
          194 References
          View all (32) Claudin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 1 Polyclonal Antibody (MH25)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (88)
          • Advanced Verification (2)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 94

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 1 Antibody (71-7800) in IHC (P)

          Immunohistochemistry analysis of Claudin-1 showing staining in the membrane and weak cytoplasm staining of paraffin-embedded human liver tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          Claudin 1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody
          Claudin 1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Claudin 1 Polyclonal Antibody (MH25)

          Product Details

          71-7800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 80 publications 80 publications

          Immunohistochemistry (IHC)

          -
          View 46 publications 46 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          View 8 publications 8 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          -
          View 46 publications 46 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Avian, Bovine, Chicken, Dog, Fish, Human, Mouse, Pig, Rabbit, Rat, Tag, Virus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Clone

          MH25

          Immunogen

          A synthetic peptide derived from the C-terminus of the human/mouse Claudin-1 protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533997

          Product Specific Information

          This antibody reacts with the ~ 22 kDa Claudin-1 protein. Reactivity was confirmed by western blotting and immunofluorescence. Positive controls include MDCK cells (dog), Caco-2cells (human), mouse liver, rat liver, and rat kidney.

          NOTE: This antibody strongly cross reacts with the Claudin-3 protein. It has been observed in western blotting inhibition studies using Caco-2 cell lysates, that this antibody is inhibited by addition of Claudin-3 peptide. For a Claudin-1 specific antibody that does not cross-react with Claudin-3, please order Cat. No. 519000 (Rb x Claudin-1) instead.

          Target Information

          Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CLDN1; senescence-associated epithelial membrane protein 1; tight junction transmembrane protein; unnamed protein product

          View more View less

          Gene Aliases: CLD1; ILVASC; SEMP1

          View more View less

          UniProt ID: (Human) O95832, (Mouse) O88551, (Rat) P56745

          View more View less

          Entrez Gene ID: (Human) 9076, (Dog) 608207, (Mouse) 12737, (Rat) 65129

          View more View less

          Function(s)
          virus receptor activity structural molecule activity protein binding identical protein binding tight junction
          Process(es)
          establishment of blood-nerve barrier response to toxic substance calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules positive regulation of cell migration response to lipopolysaccharide cell junction maintenance maintenance of blood-brain barrier cell-cell junction organization response to ethanol viral entry into host cell protein oligomerization positive regulation of epithelial cell proliferation involved in wound healing establishment of skin barrier xenobiotic transport across blood-nerve barrier response to interleukin-18 bicellular tight junction assembly cellular response to lead ion cellular response to interferon-gamma cellular response to tumor necrosis factor response to dexamethasone cellular response to transforming growth factor beta stimulus positive regulation of wound healing establishment of endothelial intestinal barrier liver regeneration positive regulation of bicellular tight junction assembly cellular response to butyrate
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch