Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Sodium Channel Pan Antibodies

          Antibodies Inc

          Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40)

          View all (17) Sodium Channel Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Pan-Nav1 Sodium Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Pan-Nav1 Sodium Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan-Nav1 Sodium Channel Antibody (75-405) in IHC (F)

          Immunofluorescence staining of adult rat cortex with N419/40 (red), K89/34 (green) and Hoechst nuclear stain (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan-Nav1 Sodium Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Pan-Nav1 Sodium Channel Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40)

          Product Details

          75-405

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:250
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N419/40

          Immunogen

          Synthetic peptide amino acids 1501-1518 of human Nav1.1 (accession number P35498).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Sequence of this protein is as follows: TEEQKKYYNA MKKLGSKK

          Target Information

          Voltage-gated sodium channels (NaV) are essential for the generation of action potentials and for cell excitability. NaV channels are activated in response to depolarization and selectively allow flow of Na+ ions. To date, nine NaV alpha subunits have been cloned and named NaV1.1-NaV1.9. The NaV channels are classified into two groups according to their sensitivity to tetrodotoxin (TTX): TTX-sensitive (NaV1.1, NaV1.2, NaV1.3, NaV1.4, NaV1.6 and NaV1.7) and TTX-resistant (NaV1.5, NaV1.8 and NaV1.9). Mammalian sodium channels are heterotrimers, composed of a central, pore-forming alpha subunit and two auxiliary beta subunits. The expression of the alpha subunit isoform is developmentally regulated and tissue specific. Sodium channels in the adult central nervous system and heart contain beta 1 through beta 4 subunits, whereas sodium channels in adult skeletal muscle have only the beta 1 subunit.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3-Feb; ataxia 3; brain III voltage-gated sodium channel; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CTC-264K15.6; H1B; hH1c; voltage-gated sodium channel; hNa6/Scn8a voltage-gated sodium channel; hNE-Na; mu-1; Na+ channel; naCh6; NaCh6; Scn8a; NaN alpha subunit; Nav1.4; Nav1.5/SCN5A.3; NaV1.8; neuroendocrine sodium channel; peripheral nerve protein type 4; peripheral nerve sodium channel 3; peripheral nerve sodium channel 5; peripheral sodium channel 1; PN4; rCG26412-like; RIIA sodium channel protein; SCN10A/PN3; scn5a-JEJ, HH1, hNav1.5, LQT3; SCN8A; sensory neuron sodium channel; sensory neuron sodium channel 2; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; skeletal muscle voltage-sensitive sodium channel subtype 2; skM1; sodium channel 25; sodium channel 6; sodium channel alpha-subunit; sodium channel protein brain I subunit alpha; sodium channel protein brain II subunit alpha; sodium channel protein brain III subunit alpha; sodium channel protein cardiac muscle subunit alpha; sodium channel protein skeletal muscle subunit alpha; sodium channel protein type 1 subunit alpha-like; sodium channel protein type 3 subunit alpha-like; sodium channel protein type I subunit alpha; sodium channel protein type II subunit alpha; sodium channel protein type III subunit alpha; sodium channel protein type IV subunit alpha; sodium channel protein type IX subunit alpha; sodium channel protein type V subunit alpha; sodium channel protein type VIII subunit alpha; sodium channel protein type X subunit alpha; sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type IX alpha; sodium channel voltage-gated type VIII alpha; sodium channel voltage-gated type X alpha polypeptide; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage-gated, type 1, alpha polypeptide; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type 2, alpha 1 polypeptide; sodium channel, voltage-gated, type 2, alpha 1 subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type 4, alpha polypeptide; sodium channel, voltage-gated, type 4, alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type 8, alpha polypeptide; sodium channel, voltage-gated, type 8, alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type I, alpha polypeptide; sodium channel, voltage-gated, type I, alpha subunit; sodium channel, voltage-gated, type II, alpha 1; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha polypeptide 1; sodium channel, voltage-gated, type II, alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type III, alpha subunit; sodium channel, voltage-gated, type IV, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium channel, voltage-gated, type VIII, alpha polypeptide; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium channel, voltage-gated, type XI, alpha; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; sodium ion channel; SP-19; splice variant; TTX-R sodium channel; unnamed protein product; voltage gated sodium channel; voltage gated sodium channel alpha subunit; voltage-dependent sodium channel; voltage-gated sodium channel alpha subunit; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subtype II; voltage-gated sodium channel subtype III; voltage-gated sodium channel subunit alpha Nav1.1; voltage-gated sodium channel subunit alpha Nav1.2; voltage-gated sodium channel subunit alpha Nav1.3; voltage-gated sodium channel subunit alpha Nav1.4; voltage-gated sodium channel subunit alpha Nav1.5; voltage-gated sodium channel subunit alpha Nav1.6; voltage-gated sodium channel subunit alpha Nav1.7; voltage-gated sodium channel subunit alpha Nav1.8; voltage-gated sodium channel subunit alpha Nav1.9; voltage-gated sodium channel type II alpha subunit; voltage-gated sodium channel type VIII alpha protein; voltage-gated sodium channel; alpha subunit domain 4, S5-S6; voltage-sensitive sodium channel alpha subunit

          View more View less

          Gene Aliases: 6430408L10; A230052E19Rik; B230332M13; BFIC3; BFIS3; BFIS5; BFNIS; C630029C19Rik; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; CMYO22A; CMYP22A; DEE11; DEE13; DEE6; DEE62; DEE6A; DEE6B; dmu; DRVT; EA9; EIEE11; EIEE13; EIEE6; EIEE62; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FFEVF4; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSCI; HBSCII; HH1; HOKPP2; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; LQT3; MED; mH1; mH2; microI; mKIAA4197; mnd-2; mnd2; MYOCL2; Na(v)1.2; Na(V)1.4; Na(V)1.8; NAC1; NAC1A; NAC2; NAC3; NaCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NAV1.9; NCHVS; NE-NA; NENA; nmf2; nmf335; nmf58; NSS2; nur14; PFHB1; PN1; PN3; PN4; PN5; RATRSKM2X; RII/RIIA; RNSCPIIR; rSkM2; RSKM2X; SCAL; SCIII; SCN; SCN1; SCN12A; Scn2a; SCN2A1; SCN2A2; Scn2x; ScpII; seal; SFNP; SkM1; SkM2; SMEI; SNS; SNS-2; SNS2; SSS1; VF1

          View more View less

          UniProt ID: (Human) Q9Y5Y9, (Human) Q9UI33, (Human) P35498, (Human) Q99250, (Human) Q9NY46, (Human) P35499, (Human) Q14524, (Human) Q9UQD0, (Human) Q15858, (Mouse) Q6QIY3, (Rat) Q62968, (Mouse) Q9R053, (Rat) O88457, (Rat) P04774, (Mouse) A2APX8, (Rat) P04775, (Mouse) B1AWN6, (Rat) P08104, (Mouse) A2ASI5, (Rat) P15390, (Mouse) Q9ER60, (Rat) P15389, (Mouse) Q9JJV9, (Mouse) Q9WTU3, (Rat) O88420, (Rat) O08562, (Mouse) Q62205

          View more View less

          Entrez Gene ID: (Human) 6336, (Human) 11280, (Human) 6323, (Human) 6326, (Human) 6328, (Human) 6329, (Human) 6331, (Human) 6334, (Human) 6335, (Mouse) 20264, (Rat) 29571, (Mouse) 24046, (Rat) 29701, (Rat) 81574, (Mouse) 20265, (Rat) 24766, (Mouse) 110876, (Rat) 497770, (Mouse) 20269, (Rat) 25722, (Mouse) 110880, (Rat) 25665, (Mouse) 20271, (Mouse) 20273, (Rat) 29710, (Rat) 78956, (Mouse) 20274

          View more View less

          Function(s)
          ion channel activity voltage-gated sodium channel activity cation channel activity ion channel binding voltage-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential protein binding calmodulin binding fibroblast growth factor binding enzyme binding protein kinase binding protein domain specific binding ankyrin binding ubiquitin protein ligase binding nitric-oxide synthase binding voltage-gated sodium channel activity involved in cardiac muscle cell action potential voltage-gated sodium channel activity involved in AV node cell action potential voltage-gated sodium channel activity involved in bundle of His cell action potential voltage-gated sodium channel activity involved in Purkinje myocyte action potential voltage-gated sodium channel activity involved in SA node cell action potential scaffold protein binding sodium ion binding sodium channel activity leucine zipper domain binding
          Process(es)
          action potential ion transport sodium ion transport nervous system development memory intrinsic apoptotic signaling pathway in response to osmotic stress neuronal action potential sodium ion transmembrane transport myelination neuron apoptotic process transmembrane transport cellular response to hypoxia cardiac muscle cell action potential involved in contraction peripheral nervous system development membrane depolarization during action potential muscle organ development sensory perception of sound locomotory behavior adult walking behavior response to toxic substance sensory perception of pain nerve development regulation of heart rate cardiac ventricle development brainstem development positive regulation of heart rate positive regulation of sodium ion transport response to denervation involved in regulation of muscle adaptation telencephalon development cerebellum development odontogenesis of dentin-containing tooth positive regulation of action potential animal organ development system development positive regulation of epithelial cell proliferation membrane depolarization cardiac muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane depolarization regulation of atrial cardiac muscle cell membrane repolarization regulation of ventricular cardiac muscle cell membrane depolarization cellular response to calcium ion regulation of cardiac muscle cell contraction ventricular cardiac muscle cell action potential membrane depolarization during cardiac muscle cell action potential atrial cardiac muscle cell action potential SA node cell action potential AV node cell action potential bundle of His cell action potential membrane depolarization during AV node cell action potential membrane depolarization during SA node cell action potential membrane depolarization during Purkinje myocyte cell action potential membrane depolarization during bundle of His cell action potential cell communication involved in cardiac conduction AV node cell to bundle of His cell communication regulation of heart rate by cardiac conduction regulation of muscle system process sodium ion import across plasma membrane membrane depolarization during atrial cardiac muscle cell action potential regulation of sodium ion transmembrane transport determination of adult lifespan neuronal action potential propagation regulation of membrane potential neuromuscular process controlling posture detection of mechanical stimulus involved in sensory perception of pain regulation of presynaptic membrane potential thigmotaxis acute inflammatory response chronic inflammatory response inflammatory response G-protein coupled receptor signaling pathway axonogenesis circadian rhythm chemosensory behavior response to heat response to cold response to xenobiotic stimulus response to high light intensity response to auditory stimulus transmission of nerve impulse response to prostaglandin E thermosensory behavior mast cell degranulation response to pain behavioral response to pain cell motility detection of temperature stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception reflex micturition skeletal muscle organ development artery development behavioral response to acetic acid induced pain behavioral response to formalin induced pain cellular response to cold calcium ion transmembrane transport response to nitric oxide action potential initiation cAMP/PKA signal transduction sensory perception of itch calcitonin gene-related peptide receptor signaling pathway small intestine smooth muscle contraction regulation of ion transmembrane transport regulation of cardiac muscle contraction cardiac conduction dentate gyrus development optic nerve development cerebral cortex development corpus callosum development axon development response to pyrethroid cellular response to antibiotic cation transport potassium ion transport choline transport regulation of skeletal muscle contraction by action potential post-embryonic development rhythmic excitation negative regulation of action potential action potential propagation sensory perception cardiac conduction system development muscle contraction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline