RediLoad™ is a loading buffer designed for easy loading and tracking of PCR products run in agarose gels. It isRead more
Have Questions?
Catalog Number
Quantity
750026
10 mL
Catalog number 750026
Price (USD)
242.65
Online exclusive
261.00
Save 18.35
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
242.65
Online exclusive
261.00
Save 18.35
Each
RediLoad™ is a loading buffer designed for easy loading and tracking of PCR products run in agarose gels. It is added directly to the PCR reaction mixture prior to amplification. RediLoad™ features a reagent that increases density for easy loading, and a red tracking dye that migrates at approximately 25 bp and allows easy monitoring of electrophoresis. RediLoad™ can be used as a replacement for conventional agarose loading buffer and is supplied in a 10X concentration.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration10X
Gel CompatibilityE-Gel™, Agarose Gels
Quantity10 mL
Shipping ConditionRoom Temperature
Gel TypeAgarose
Product LineReady-Load
Product TypeLoading Buffer
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
Do you offer standalone gel loading dyes that can be added to a PCR reaction, before the reaction?
We offer the following red dye, RediLoad Loading Buffer (Cat. No. 750026).