Do you offer standalone gel loading dyes that can be added to a PCR reaction, before the reaction?
We offer the following red dye, RediLoad Loading Buffer (Cat. No. 750026).
Search
View additional product information for RediLoad™ Loading Buffer - FAQs (750026)
1 product FAQs found