Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Search
View additional product information for 2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge - FAQs (75003872, 75003871, 75003870)
1 product FAQs found