mySPIN 6 Mini Centrifuge with two Rotors, Storage Case, Adapters, Tube Rack, Universal Power Adapter - FAQs
mySPIN 6 Mini Centrifuge with two Rotors, Storage Case, Adapters, Tube Rack, Universal Power Adapter - FAQs
View additional product information for mySPIN™ 6 Mini Centrifuge - FAQs (75004061, 75004086, 75004087)
1 product FAQs found
Does the mySPIN 6 Mini Centrifuge come with a rotor?
The mySPIN 6 Mini Centrifuge includes 2 rotors and adapters to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes.