beta-Galactosidase Assay Reagent
Thermo Scientific™

beta-Galactosidase Assay Reagent

Lyse cultured mammalian cells and measure beta-galactosidase activity using compatible M-PER Cell Lysis Reagent and colorimetric b-Gal Assay Reagent.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7570525 mL
757103 x 25 mL
Catalog number 75705
Price (USD)
169.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
169.00
Each

The Thermo Scientific Mammalian β-Galactosidase Assay Reagent is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Features of the Thermo Scientific Mammalian β-Galactosidase Assay Kit (assay reagent and stop solution):

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatible with prepared lysates from other species such as bacteria

Related Products

Mammalian beta-Galactosidase Assay Kit (Cat. No. 75707)

beta-Galactosidase Assay Stop Solution (Cat. No. 75706)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayReporter Enzyme
For Use With (Application)Absorbance
For Use With (Equipment)Microplate Reader
Format96-well Plate
Label or DyeEnzyme Labeled
Product LinePierce
Product TypePierce beta-Galactosidase Assay Reagent
Quantity25 mL
SubstrateBeta-Gal Substrate
Detection MethodAbsorbance
FormLiquid
Substrate PropertiesChemical Substrate
Target EnzymeBeta-Galactosidase
Unit SizeEach
Contents & Storage
Store at -20°C.