Reduced glutathione dry powder sufficient for preparing 6 mL of 0.1M glutathione solution.Related ProductsPierce™ Glutathione AgarosePierce™ Glutathione Superflow AgaroseRead more
Have Questions?
Catalog Number
Quantity
78259
5 x 184 mg
Catalog number 78259
Price (USD)
89.65
Online exclusive
102.00
Save 12.35
Each
Quantity:
5 x 184 mg
Request bulk or custom format
Price (USD)
89.65
Online exclusive
102.00
Save 12.35
Each
Reduced glutathione dry powder sufficient for preparing 6 mL of 0.1M glutathione solution.