Pierce™ Glutathione Magnetic Agarose Beads
Thermo Scientific™

Pierce™ Glutathione Magnetic Agarose Beads

Thermo Scientific™ Pierce™ Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST) from a bacterial,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7860220 mL
786014 mL
Catalog number 78602
Price (USD)
724.65
Online exclusive
814.00
Save 89.35
Each
Quantity:
20 mL
Price (USD)
724.65
Online exclusive
814.00
Save 89.35
Each

Thermo Scientific™ Pierce™ Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST) from a bacterial, yeast, or mammalian crude cell lysate. Pierce™ Glutathione Magnetic Agarose Beads are well suited for purifying GST fusion proteins from a soluble protein extract and are ideal for the purification of proteins expressed at low levels from diluted supernatants. The beads can be used in manual applications with a magnetic stand or automated applications with an instrument such as the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purification and screening of purification conditions.

Download the KingFisher™ Duo BindIt™ Software protocol (.bdz) >

Download the KingFisher™ Flex BindIt™ Software protocol (.bdz) >

Features of Glutathione Magnetic Agarose Beads:

  • High-performance beads—non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS applications
  • Stable affinity ligand—glutathione is covalently immobilized to particles, enabling leach-resistance and clean purification products
  • High capacity—binding capacity is sufficient for both routine and demanding purification procedures

The high-performance, magnetite, superparamagnetic particles are validated and optimized for use with high-throughput magnetic platforms, such as the KingFisher™ 96 and KingFisher™ Flex magnetic particle processors, but the beads also enable premium performance for simple benchtop purification applications using an appropriate magnetic stand.

Glutathione S-transferase (GST) is a protein that is commonly added to the end of recombinant proteins to aid in their purification or detection. To make an affinity support, glutathione is immobilized through its central carbon, preserving the essential structure necessary for efficient GST binding. Addition of reduced glutathione in a physiologic buffer is sufficient to competitively displace and recover the GST fusion protein from the immobilized glutathione.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume5 mL
ConcentrationSlurry: 25%
FormatBottle
Quantity20 mL
Shipping ConditionWet Ice
Stationary PhaseGlutathione
Capacity (Metric)20 mL
FormLiquid Suspension
Particle Size30 μm
Product LinePierce
TypeMagnetic Agarose Bead
Unit SizeEach
Contents & Storage
5 mL settled resin. Supplied as 20 mL of 25% v/v suspension in 20% ethanol.

Store at 2–8°C.

Frequently asked questions (FAQs)

Can the Pierce Magnetic Agarose Beads be frozen?

No. Freezing will adversely affect the agarose structure and may cause bead aggregation or loss of binding.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can the Pierce Magnetic Agarose Beads be used in an automated magnetic particle processor?

Yes. Protocols for using the beads on a KingFisher instrument are available on the product page.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What advantage do Pierce Magnetic Agarose beads have over Dynabeads or Pierce Magnetic Beads?

Pierce Magentic Agarose Beads have a much higher binding capacity.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (3)

Citations & References
Abstract
Phospho-regulation of the Shugoshin-Condensin interaction at the centromere in budding yeast
Authors:Metwaly GY, Wu Y, Peplowska K, Rohri J, Soh Y-M, Burmann F, Gruber S, Storchova Z
Journal:PLoS Genet
PubMed ID:
A novel family of expression vectors with multiple affinity tags for wheat germ cell-free protein expression
Authors:Nagy SK, Kallai BM, Andras J, Meszaros T
Journal:BMC Biotechnology
PubMed ID:
pUL21 is a viral phosphatase adaptor that promotes herpes simplex virus replication and spread
Authors:Benedyk TH, Muenzner J, Connor V, Han Y, Brown K, Wijesinghe KJ, Zhuang Y, Colaco S, Stoll GA, Tutt OS, Svobodova S, Svergun DI, Bryant NA, Deane JE, Firth AE, Jeffries CM, Crump CM, Graham SC
Journal:PLoS Pathog
PubMed ID: